Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEKNPPDDTG PVHVPLGHIV ANEKWRGSQL AQEMQGKIKL IFEDGLTPDF YLSNRCCILY VTEADLVAGN GYRKRLVRVR NSNNLKGIVV VEKTRMSEQY FPALQKFTVL DLGMVLLPVA SQMEASCLVI QLVQEQTKEP SKNPLLGKKR ALLLSEPSLL RTVQQIPGVG KVKAPLLLQK FPSIQQLSNA SIGELEQVVG QAVAQQIHAF FTQPR
FAAP24 is a component of the Fanconi anemia (FA) core complex (see MIM 227650), which plays a crucial role in DNA damage response (Ciccia et al., 2007 [PubMed 17289582]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fanconi anemia core complex associated protein 24; Fanconi anemia core complex-associated protein 24; Fanconi anemia-associated protein of 24 kDa; unnamed protein product
Gene Aliases: C19orf40; FAAP24
UniProt ID: (Human) Q9BTP7
Entrez Gene ID: (Human) 91442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support