Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Complement C3a/C3b/C3c Antibodies

          Proteintech

          C3/C3b/C3c Polyclonal Antibody

          51 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite C3/C3b/C3c Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          C3/C3b/C3c Antibody (21337-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded Human liver cancer tissue slide using (Product # 21337-1-AP) C3/C3b/C3c antibody at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C3/C3b/C3c Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          C3/C3b/C3c Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          C3/C3b/C3c Antibody in Western Blot (WB)
          C3/C3b/C3c Antibody in Western Blot (WB)
          C3/C3b/C3c Antibody in Immunoprecipitation (IP)
          C3/C3b/C3c Polyclonal Antibody

          Product Details

          21337-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 25 publications 25 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 17 publications 17 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          C3/C3b/C3c Fusion Protein Ag15537 (1314-1663 aa encoded by BC150179)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Amount

          45 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.

          Immunogen sequence: ESASLLRSE ETKENEGFTV TAEGKGQGTL SVVTMYHAKA KDQLTCNKFD LKVTIKPAPE TEKRPQDAKN TMILEICTRY RGDQDATMSI LDISMMTGFA PDTDDLKQLA NGVDRYISKY ELDKAFSDRN TLIIYLDKVS HSEDDCLAFK VHQYFNVELI QPGAVKVYAY YNLEESCTRF YHPEKEDGKL NKLCRDELCR CAEENCFIQK SDDKVTLEER LDKACEPGVD YVYKTRLVKV QLSNDFDEYI MAIEQTIKSG SDEVQVGQQR TFISPIKCRE ALKLEEKKHY LMWGLSSDFW GEKPNLSYII GKDTWVEHWP EEDECQDEEN QKQCQDLGAF TESMVVFGCP N (1314-1663 aa encoded by BC150179)

          Target Information

          Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: acylation-stimulating protein cleavage product; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Complement C3; complement component 3; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; HSE-MSF; prepro-C3; unnamed protein product

          View more View less

          Gene Aliases: AHUS5; AI255234; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p; HSE-MSF; Plp

          View more View less

          UniProt ID: (Human) P01024, (Rat) P01026, (Mouse) P01027

          View more View less

          Entrez Gene ID: (Human) 718, (Rat) 24232, (Mouse) 12266

          View more View less

          Function(s)
          antigen binding endopeptidase inhibitor activity receptor binding protein binding chemokine activity C5L2 anaphylatoxin chemotactic receptor binding receptor agonist activity lipid binding cofactor binding protease inhibitor
          Process(es)
          positive regulation of type IIa hypersensitivity complement activation, lectin pathway activation of membrane attack complex positive regulation of protein phosphorylation positive regulation of activation of membrane attack complex immune system process complement receptor mediated signaling pathway lipid metabolic process fatty acid metabolic process inflammatory response immune response complement activation complement activation, alternative pathway complement activation, classical pathway signal transduction G-protein coupled receptor signaling pathway opsonization response to bacterium positive regulation of vascular endothelial growth factor production positive regulation of glucose transport regulation of triglyceride biosynthetic process positive regulation of lipid storage neuron remodeling leukocyte chemotaxis killing of cells of other organism oviduct epithelium development B cell activation innate immune response positive regulation of G-protein coupled receptor protein signaling pathway positive regulation of angiogenesis positive regulation of receptor-mediated endocytosis positive regulation of phagocytosis protein maturation positive regulation of phagocytosis, engulfment beta-amyloid clearance complement-dependent cytotoxicity complement-mediated synapse pruning vertebrate eye-specific patterning complement activation, GZMK pathway positive regulation of apoptotic cell clearance tolerance induction chemotaxis blood coagulation negative regulation of endopeptidase activity response to magnesium ion response to estradiol response to progesterone response to estrogen positive regulation of developmental growth response to glucocorticoid positive regulation of ERK1 and ERK2 cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline