Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • cGAS Antibodies

          Proteintech

          C6orf150 Polyclonal Antibody

          53 References
          View all (23) cGAS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite C6orf150 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          C6orf150 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          C6orf150 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          C6orf150 Antibody (26416-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed HeLa cells using 26416-1-AP (C6orf150 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          C6orf150 Antibody in Immunocytochemistry (ICC/IF)
          C6orf150 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          C6orf150 Antibody in Western Blot (WB)
          C6orf150 Antibody in Western Blot (WB)
          C6orf150 Antibody in Western Blot (WB)
          C6orf150 Polyclonal Antibody

          Product Details

          26416-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:750-1:3000
          View 30 publications 30 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 13 publications 13 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          C6orf150 Fusion Protein Ag23455 (1-233 aa encoded by BC113608)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MQPWHGKAM QRASEAGATA PKASARNARG APMDPNESPA APEAALPKAG KFGPARKSGS RQKKSAPDTQ ERPPVRATGA RAKKAPQRAQ DTQPSDATSA PGAEGLEPPA AREPALSRAG SCRQRGARCS TKPRPPPGPW DVPSPGLPVS APILVRRDAA PGASKLRAVL EKLKLSRDDI STAAGMVKGV VDHLLLRLKC DSAFRGVGLL NTGSYYEHVK ISAPNEFDVM FKLE (1-233 aa encoded by BC113608)

          Target Information

          cGAS (Cyclic GMP-AMP synthase) is a nucleotidyltransferase that catalyzes the formation of cyclic GMP-AMP (cGAMP) from ATP and GTP and plays a key role in innate immunity. Catalysis involves both the formation of 2', 5' phosphodiester linkage at the GpA step and the formation of 3', 5' phosphodiester linkage at the ApG step. This produces c[G(2',5')pA(3'5')p] responses. cGAS also binds cytosolic DNA directly, leading to the activation and synthesis of cGAMP. Then, a second messenger binds to and activates TMEM173/STING thereby triggering type-I interferon production. cGAS is also involved in innate immune sensorship of infection, detection of DNA from bacteria, response to cellular stress, DNA damage, and regulation of cellular senescence.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2'3'-cGAMP synthase; C6orf150; C6orf150; cGAS is a cytosolic DNA sensor that induces interferons by producing the second messenger cGAMP; cGAMP synthase; Cyclic GMP-AMP synthase; DANGER family member 4; h-cGAS; Mab-21 domain containing 1; Mab-21 domain-containing protein 1; MB21D1; protein MB21D1; unnamed protein product

          View more View less

          Gene Aliases: C6orf150; CGAS; D4; h-cGAS; MB21D1

          View more View less

          UniProt ID: (Human) Q8N884

          View more View less

          Entrez Gene ID: (Human) 115004

          View more View less

          Function(s)
          nucleotide binding DNA binding chromatin binding double-stranded DNA binding protein binding ATP binding GTP binding phosphatidylinositol-4,5-bisphosphate binding lipid binding transferase activity nucleotidyltransferase activity nucleosome binding protein homodimerization activity metal ion binding cyclic-GMP-AMP synthase activity molecular condensate scaffold activity poly-ADP-D-ribose modification-dependent protein binding
          Process(es)
          activation of innate immune response pattern recognition receptor signaling pathway positive regulation of defense response to virus by host immune system process regulation of immunoglobulin production cytoplasmic pattern recognition receptor signaling pathway DNA repair cellular response to DNA damage stimulus determination of adult lifespan cAMP-mediated signaling cGMP-mediated signaling signal transduction involved in regulation of gene expression regulation of type I interferon production positive regulation of type I interferon production paracrine signaling innate immune response negative regulation of DNA repair regulation of immune response regulation of T cell activation defense response to virus cellular response to exogenous dsRNA cGAS/STING signaling pathway negative regulation of cGAS/STING signaling pathway positive regulation of cytokine production involved in inflammatory response negative regulation of double-strand break repair via homologous recombination positive regulation of cellular senescence
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0