Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: GTSNLSETEP PLWKESPGQL SDYRVENSMY IINPWVYLER MGMYKIILNQ TARYFAKFAP DNEQNILWGL PLQYGWQYRT GRLADPTRR
C6orf58 is a protein coding gene. May be involved in early liver development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein LEG1 homolog; UPF0762 protein C6orf58
Gene Aliases: C6orf58; LEG1
UniProt ID: (Human) Q6P5S2
Entrez Gene ID: (Human) 352999
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support