Recommended positive controls: U87-MG.
Predicted reactivity: Mouse (82%), Rat (85%), Rabbit (93%), Bovine (83%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.