Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KGAILTTMLA TRNFSAAKSL LKKPDGVKES TESSNTTIED EDVKARKQEI IKVTEQLIEA INNGDFEAYT KICDPGLTAF EPEALGNLVE GMDFHRFYFE NALSKSNKPI
CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase type II delta chain; Calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II delta subunit; CaM kinase II subunit delta; CaM-kinase II delta chain; CAM2; CaMK-II delta subunit; caMK-II subunit delta; CAMK2B; CAMK2G; CAMKB; CAMKG
Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CaMK II; CAMK2D; CAMKD; Kiaa4163
UniProt ID: (Human) Q13557, (Mouse) Q6PHZ2
Entrez Gene ID: (Human) 817, (Mouse) 108058
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support