Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Aliquoting is unnecessary for -20°C storage.
Calpain, an intracellular calcium-dependent protease that cleaves cytoskeletal and submembranous proteins, plays a role in cytoskeletal reorganization and muscle protein degradation. Calpain is a heterodimer composed of a small regulatory subunit and one of three large catalytic subunits, designated calpain 1, calpain 2 and calpain 3. Calpain 3 (calpain p94) is a muscle-preferred calcium activated neutral protease that localizes to the nucleus. Mutations involving the calpain 3 gene are associated with limb-girdle muscle dystrophy type 2A, a form of autosomal recessive and progressive neuromuscular diso4. Calpastatin regulates calpain by inhibiting both the proteolytic activity of calpain and its binding to membranes. Calpastatin exists in two types, tissue type and erythrocyte type, resulting from both alternative splicing and proteolytic processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium dependent cysteine protease; calcium-activated neutral proteinase 3; calpain 3 (p94) isoform CAPN3_1; calpain 3 (p94) isoform CAPN3_2; calpain 3 (p94) isoform CAPN3_7; calpain L3; calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; CANP 3; CAPN 3; cysteine protease; hMp78; cysteine protease; hMp84; LGMD 2A; MGC10767; MGC11121; MGC14344; MGC4403; muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; new calpain 1; p94 protein; retina-specific; splice variant of the product encoded by GenBank Accession Number AF061726; splice variant; splice variant of muscle type calpain missing insert regions; unnamed protein product
Gene Aliases: CANP3; CANPL3; LGMD2; LGMD2A; LGMDD4; LGMDR1; Lp82; Lp84; Lp85; nCL-1; p94
UniProt ID: (Human) P20807, (Rat) P16259
Entrez Gene ID: (Human) 825, (Rat) 29155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support