Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Calcium Sensing Receptor Antibodies

          Abnova

          CASR Polyclonal Antibody, MaxPab™

          View all (24) Calcium Sensing Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CASR Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CASR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CASR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CASR Antibody (H00000846-B01P) in WB

          Western Blot analysis of CASR expression in transfected 293T cell line by CASR MaxPab polyclonal antibody. Lane 1: CASR transfected lysate(120.60 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CASR Antibody in Western Blot (WB)

          Product Details

          H00000846-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CASR (NP_000379.2, 1 a.a. approximately 1078 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAFYSCCWVL LALTWHTSAY GPDQRAQKKG DIILGGLFPI HFGVAAKDQD LKSRPESVEC IRYNFRGFRW LQAMIFAIEE INSSPALLPN LTLGYRIFDT CNTVSKALEA TLSFVAQNKI DSLNLDEFCN CSEHIPSTIA VVGATGSGVS TAVANLLGLF YIPQVSYASS SRLLSNKNQF KSFLRTIPND EHQATAMADI IEYFRWNWVG TIAADDDYGR PGIEKFREEA EERDICIDFS ELISQYSDEE EIQHVVEVIQ NSTAKVIVVF SSGPDLEPLI KEIVRRNITG KIWLASEAWA SSSLIAMPQY FHVVGGTIGF ALKAGQIPGF REFLKKVHPR KSVHNGFAKE FWEETFNCHL QEGAKGPLPV DTFLRGHEES GDRFSNSSTA FRPLCTGDEN ISSVETPYID YTHLRISYNV YLAVYSIAHA LQDIYTCLPG RGLFTNGSCA DIKKVEAWQV LKHLRHLNFT NNMGEQVTFD ECGDLVGNYS IINWHLSPED GSIVFKEVGY YNVYAKKGER LFINEEKILW SGFSREVPFS NCSRDCLAGT RKGIIEGEPT CCFECVECPD GEYSDETDAS ACNKCPDDFW SNENHTSCIA KEIEFLSWTE PFGIALTLFA VLGIFLTAFV LGVFIKFRNT PIVKATNREL SYLLLFSLLC CFSSSLFFIG EPQDWTCRLR QPAFGISFVL CISCILVKTN RVLLVFEAKI PTSFHRKWWG LNLQFLLVFL CTFMQIVICV IWLYTAPPSS YRNQELEDEI IFITCHEGSL MALGFLIGYT CLLAAICFFF AFKSRKLPEN FNEAKFITFS MLIFFIVWIS FIPAYASTYG KFVSAVEVIA ILAASFGLLA CIFFNKIYII LFKPSRNTIE EVRCSTAAHA FKVAARATLR RSNVSRKRSS SLGGSTGSTP SSSISSKSNS EDPFPQPERQ KQQQPLALTQ QEQQQQPLTL PQQQRSQQQP RCKQKVIFGS GTVTFSLSFD EPQKNAMAHG NSTHQNSLEA QKSSDTLTRH QPLLPLQCGE TDLDLTVQET GLQGPVGGDQ RPEVEDPEEL SPALVVSSSQ SFVISGGGST VTENVVNS

          Target Information

          Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Ca sensing receptor; calcium receptor|CaR; CaR; CaSR antibody; Extracellular calcium-sensing receptor; parathyroid Ca(2+)-sensing receptor 1; Parathyroid cell calcium-sensing receptor 1; PCaR1

          View more View less

          Gene Aliases: CAR; CASR; EIG8; FHH; FIH; GPRC2A; hCasR; HHC; HHC1; HYPOC1; NSHPT; PCAR1

          View more View less

          UniProt ID: (Human) P41180

          View more View less

          Entrez Gene ID: (Human) 846

          View more View less

          Function(s)
          G-protein coupled receptor activity integrin binding calcium ion binding protein binding amino acid binding protein kinase binding identical protein binding protein homodimerization activity ion channel binding metal ion binding
          Process(es)
          ossification response to ischemia detection of calcium ion cellular calcium ion homeostasis signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway JNK cascade chemosensory behavior positive regulation of cell proliferation anatomical structure morphogenesis response to metal ion positive regulation of gene expression positive regulation of insulin secretion bile acid secretion cellular response to hepatocyte growth factor stimulus vasodilation positive regulation of vasoconstriction branching morphogenesis of an epithelial tube positive regulation of positive chemotaxis response to calcium ion regulation of calcium ion transport fat pad development positive regulation of ERK1 and ERK2 cascade cellular response to vitamin D cellular response to glucose stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hypoxia response to fibroblast growth factor positive regulation of calcium ion import regulation of presynaptic membrane potential adenylate cyclase-activating G protein-coupled cAMP receptor signaling pathway positive regulation of NLRP3 inflammasome complex assembly cellular response to peptide chloride transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline