Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CC2D1A Antibodies

          Abnova

          CC2D1A Polyclonal Antibody, MaxPab™

          View all (18) CC2D1A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CC2D1A Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          CC2D1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CC2D1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CC2D1A Antibody (H00054862-B01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to CC2D1A on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CC2D1A Antibody in Immunocytochemistry (ICC/IF)
          CC2D1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CC2D1A Antibody in Western Blot (WB)
          CC2D1A Antibody in Western Blot (WB)
          CC2D1A Antibody in Western Blot (WB)
          CC2D1A Polyclonal Antibody, MaxPab™

          Product Details

          H00054862-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          3 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CC2D1A (NP_060191.3, 1 a.a. approximately 951 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MHKRKGPPGP PGRGAAAARQ LGLLVDLSPD GLMIPEDGAN DEELEAEFLA LVGGQPPALE KLKGKGPLPM EAIEKMASLC MRDPDEDEEE GTDEDDLEAD DDLLAELNEV LGEEQKASET PPPVAQPKPE APHPGLETTL QERLALYQTA IESARQAGDS AKMRRYDRGL KTLENLLASI RKGNAIDEAD IPPPVAIGKG PASTPTYSPA PTQPAPRIAS APEPRVTLEG PSATAPASSP GLAKPQMPPG PCSPGPLAQL QSRQRDYKLA ALHAKQQGDT TAAARHFRVA KSFDAVLEAL SRGEPVDLSC LPPPPDQLPP DPPSPPSQPP TPATAPSTTE VPPPPRTLLE ALEQRMERYQ VAAAQAKSKG DQRKARMHER IVKQYQDAIR AHKAGRAVDV AELPVPPGFP PIQGLEATKP TQQSLVGVLE TAMKLANQDE GPEDEEDEVP KKQNSPVAPT AQPKAPPSRT PQSGSAPTAK APPKATSTRA QQQLAFLEGR KKQLLQAALR AKQKNDVEGA KMHLRQAKGL EPMLEASRNG LPVDITKVPP APVNKDDFAL VQRPGPGLSQ EAARRYGELT KLIRQQHEMC LNHSNQFTQL GNITETTKFE KLAEDCKRSM DILKQAFVRG LPTPTARFEQ RTFSVIKIFP DLSSNDMLLF IVKGINLPTP PGLSPGDLDV FVRFDFPYPN VEEAQKDKTS VIKNTDSPEF KEQFKLCINR SHRGFRRAIQ TKGIKFEVVH KGGLFKTDRV LGTAQLKLDA LEIACEVREI LEVLDGRRPT GGRLEVMVRI REPLTAQQLE TTTERWLVID PVPAAVPTQV AGPKGKAPPV PAPARESGNR SARPLHSLSV LAFDQERLER KILALRQARR PVPPEVAQQY QDIMQRSQWQ RAQLEQGGVG IRREYAAQLE RQLQFYTEAA RRLGNDGSRD AAKEALYRRN LVESELQRLR R

          Target Information

          This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Akt kinase-interacting protein 1; Coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; lethal (2) giant discs homolog 2; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein; TBK1-associated protein in endolysosomes; unnamed protein product

          View more View less

          Gene Aliases: Aki-1; AKI1; CC2D1A; FREUD-1; Freud-1/Aki1; LGD2; MRT3; TAPE

          View more View less

          UniProt ID: (Human) Q6P1N0

          View more View less

          Entrez Gene ID: (Human) 54862

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding protein binding cadherin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of respiratory gaseous exchange by neurological system process regulation of transcription from RNA polymerase II promoter endosome organization positive regulation of I-kappaB kinase/NF-kappaB signaling regulation of respiratory gaseous exchange dendrite arborization apical dendrite arborization regulation of postsynapse assembly negative regulation of snRNA transcription by RNA polymerase II
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline