Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CCL5 (RANTES) Antibodies

          Invitrogen

          CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          3 Published Figures
          13 References
          View all (30) CCL5 (RANTES) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (3)
          CCL5 (RANTES) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CCL5 (RANTES) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CCL5 (RANTES) Antibody (500-P118-100UG) in WB

          Western Blot: To detect Murine RANTES (CCL5) by Western Blot analysis CCL5 (RANTES) Polyclonal Antibody (Product # 500-P118-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine RANTES (CCL5) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CCL5 (RANTES) Antibody in Western Blot (WB)
          CCL5 (RANTES) Antibody in Western Blot (WB)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in Immunohistochemistry (IHC)
          CCL5 (RANTES) Antibody in Immunohistochemistry (IHC)
          CCL5 (RANTES) Antibody in Immunohistochemistry (IHC)
          CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          Product Details

          500-P118-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.5-2.0 µg/mL
          View 4 publications 4 publications

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications

          Immunostaining (IS)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse, Rat

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Murine RANTES (CCL5)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929300

          Product Specific Information

          AA Sequence of recombinant protein: SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine RANTES (CCL5). Anti-Murine RANTES (CCL5)-specific antibody was purified by affinity chromatography employing an immobilized Murine RANTES (CCL5) matrix.

          Sandwich ELISA: To detect Murine RANTES (CCL5) by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine RANTES (CCL5) (500-P118Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine RANTES (CCL5).

          Western Blot: To detect Murine RANTES (CCL5) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine RANTES (CCL5) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-C motif chemokine 5; MGC17164; MuRantes; OTTHUMP00000197106; SIS δ; SIS-delta; SISδ; small inducible cytokine A5; Small-inducible cytokine A5; T-cell-specific protein RANTES

          View more View less

          Gene Aliases: Ccl5; MuRantes; RANTES; Scya5; SISd; TCP228

          View more View less

          UniProt ID: (Mouse) P30882

          View more View less

          Entrez Gene ID: (Mouse) 20304

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity protein kinase activity cytokine activity chemokine activity heparin binding phospholipase activator activity receptor signaling protein tyrosine kinase activator activity CCR1 chemokine receptor binding CCR5 chemokine receptor binding chemoattractant activity chemokine receptor binding protein homodimerization activity protein self-association chemokine receptor antagonist activity cytokine intercellular signal molecule
          Process(es)
          MAPK cascade positive regulation of defense response to virus by host calcium ion transport cellular calcium ion homeostasis exocytosis chemotaxis inflammatory response immune response leukocyte cell-cell adhesion G-protein coupled receptor signaling pathway cell-cell signaling response to toxic substance positive regulation of gene expression positive regulation of macrophage chemotaxis positive regulation of T cell chemotaxis positive regulation of phosphatidylinositol 3-kinase signaling positive regulation of smooth muscle cell migration positive regulation of cell migration pseudopodium assembly positive regulation of cellular biosynthetic process activation of phospholipase D activity positive regulation of fever generation lipopolysaccharide-mediated signaling pathway positive regulation of cell-cell adhesion mediated by integrin response to cytokine positive regulation of homotypic cell-cell adhesion response to interferon-gamma response to tumor necrosis factor T-helper 1 cell diapedesis positive regulation of T cell proliferation neutrophil activation positive regulation of phosphorylation positive regulation of tyrosine phosphorylation of STAT protein protein kinase B signaling cellular protein complex assembly negative regulation by host of viral transcription negative regulation of viral genome replication positive regulation of neuron differentiation positive regulation of osteoclast differentiation negative regulation of G-protein coupled receptor protein signaling pathway positive regulation of angiogenesis positive regulation of cell adhesion eosinophil chemotaxis lymphocyte chemotaxis positive regulation of smooth muscle cell proliferation positive regulation of epithelial cell proliferation regulation of insulin secretion regulation of T cell activation positive chemotaxis protein tetramerization positive regulation of calcium ion transport negative regulation of cell death positive regulation of mast cell chemotaxis positive regulation of protein tyrosine kinase activity chemokine-mediated signaling pathway negative regulation of chemokine-mediated signaling pathway negative regulation of T cell apoptotic process positive regulation of T cell apoptotic process cellular response to organic cyclic compound positive regulation of monocyte chemotaxis extracellular negative regulation of signal transduction regulation of neuron death negative regulation of neuron death negative regulation of macrophage apoptotic process negative regulation of receptor activity positive regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of T cell migration positive regulation of natural killer cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline