Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAAAAAAAGA AGSAAPAAAA GAPGSGGAPS GSQGVLIGDR LYSGVLITLE NCLLPDDKLR FTPSMSSGLD SDTETDLRVV GCELIQAAGI LLRLPQVAMA TGQVLFQRFF YTKSFVKHSM EHVSMACVHL ASKIEEAPRR IRDVINVFHR LRQLRDKKKP VPLLLDQDYV NLKNQIIKAE RRVLKELGFC VHVKHPHKII VMYLQVLECE RNQHLVQTSW
CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin Ania-6b; cyclin M; cyclin S; Cyclin-L2; paneth cell enhanced expression; Paneth cell-enhanced expression protein; PCEE; unnamed protein product
Gene Aliases: 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; ANIA-6B; Ania6b; AW261738; CCNL2; CCNM; CCNS; HCLA-150; HCLA-ISO; HLA-ISO; MNCb-5160; PCEE; SB138
UniProt ID: (Human) Q96S94, (Mouse) Q9JJA7, (Rat) Q5I0H5
Entrez Gene ID: (Human) 81669, (Mouse) 56036, (Rat) 298686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support