Recommended positive controls: Daudi.
Predicted reactivity: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCRN4L is highly similar to Nocturnin, identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4).