Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CD109 Antibodies

          Abnova

          CD109 Polyclonal Antibody, MaxPab™

          View all (18) CD109 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD109 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD109 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD109 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD109 Antibody (H00135228-B01P) in WB

          Western Blot analysis of CD109 expression in transfected 293T cell line by CD109 MaxPab polyclonal antibody. Lane 1: CD109 transfected lysate(158.95 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD109 Antibody in Western Blot (WB)
          CD109 Polyclonal Antibody, MaxPab™

          Product Details

          H00135228-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CD109 (AAI52997.1, 1 a.a. approximately 1445 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQGPPLLTAA HLLCVCTAAL AVAPGPRFLV TAPGIIRPGG NVTIGVELLE HCPSQVTVKA ELLKTASNLT VSVLEAEGVF EKGSFKTLTL PSLPLNSADE IYELRVTGRT QDEILFSNST RLSFETKRIS VFIQTDKALY KPKQEVKFRI VTLFSDFKPY KTSLNILIKD PKSNLIQQWL SQQSDLGVIS KTFQLSSHPI LGDWSIQVQV NDQTYYQSFQ VSEYVLPKFE VTLQTPLYCS MNSKHLNGTI TAKYTYGKPV KGDVTLTFLP LSFWGKKKNI TKTFKINGSA NFSFNDEEMK NVMDSSNGLS EYLDLSSPGP VEILTTVTES VTGISRNVST NVFFKQHDYI IEFFDYTTVL KPSLNFTATV KVTRADGNQL TLEERRNNVV ITVTQRNYTE YWSGSNSGNQ KMEAVQKINY TVPQSGTFKI EFPILEDSSE LQLKAYFLGS KSSMAVHSLF KSPSKTYIQL KTRDENIKVG SPFELVVSGN KRLKELSYMV VSRGQLVAVG KQNSTMFSLT PENSWTPKAC VIVYYIEDDG EIISDVLKIP VQLVFKNKIK LYWSKVKAEP SEKVSLRISV TQPDSIVGIV AVDKSVNLMN ASNDITMENV VHELELYNTG YYLGMFMNSF AVFQECGLWV LTDANLTKDY IDGVYDNAEY AERFMEENEG HIVDIHDFSL GSSPHVRKHF PETWIWLDTN MGYRIYQEFE VTVPDSITSW VATGFVISED LGLGLTTTPV ELQAFQPFFI FLNLPYSVIR GEEFALEITI FNYLKDATEV KVIIEKSDKF DILMTSNEIN ATGHQQTLLV PSEDGATVLF PIRPTHLGEI PITVTALSPT ASDAVTQMIL VKAEGIEKSY SQSILLDLTD NRLQSTLKTL SFSFPPNTVT GSERVQITAI GDVLGPSING LASLIRMPYG CGEQNMINFA PNIYILDYLT KKKQLTDNLK EKALSFMRQG YQRELLYQRE DGSFSAFGNY DPSGSTWLSA FVLRCFLEAD PYIDIDQNVL HRTYTWLKGH QKSNGEFWDP GRVIHSELQG GNKSPVTLTA YIVTSLLGYR KYQPNIDVQE SIHFLESEFS RGISDNYTLA LITYALSSVG SPKAKEALNM LTWRAEQEGG MQFWVSSESK LSDSWQPRSL DIEVAAYALL SHFLQFQTSE GIPIMRWLSR QRNSLGGFAS TQDTTVALKA LSEFAALMNT ERTNIQVTVT GPSSPSPVKF LIDTHNRLLL QTAELAVVQP TAVNISANGF GFAICQLNVV YNVKASGSSR RRRSIQNQEA FDLDVAVKEN KDDLNHVDLN VCTSFSGPGR SGMALMEVNL LSGFMVPSEA ISLSETVKKV EYDHGKLNLY LDSVNETQFC VNIPAVRNFK VSNTQDASVS IVDYYEPRRQ AVRSYNSEVK LSSCDLCSDV QGCRPCEDGA SGSHHHSSVI FIFCFKLLYF MELWL

          Target Information

          CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 150 kDa TGF-beta-1-binding protein; activated T-cell marker CD109; AMCOM; alpha2-macroglobulin/complement; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; CD109; CD109 antigen; Gov platelet alloantigens; GPI-anchored membrane protein; p180; Platelet-specific Gov antigen; r150; RP11-525G3.1; unnamed protein product

          View more View less

          Gene Aliases: CD109; CPAMD7; p180; r150

          View more View less

          UniProt ID: (Human) Q6YHK3

          View more View less

          Entrez Gene ID: (Human) 135228

          View more View less

          Function(s)
          endopeptidase inhibitor activity serine-type endopeptidase inhibitor activity peptidase inhibitor activity
          Process(es)
          hair follicle development negative regulation of keratinocyte proliferation negative regulation of transforming growth factor beta receptor signaling pathway keratinocyte proliferation regulation of keratinocyte differentiation negative regulation of wound healing stem cell proliferation osteoclast fusion negative regulation of stem cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline