Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: QDSRGCARWC PQNSSCVNAT ACRCNPGFSS FSEIITTPTE TCDDINECAT PSKVSCGKFS DCWNTEGSYD CVCSPGYEPV SGAKTFKNES ENTCQDVDEC SSGQHQCDSS TVCFNTVGSY SCRCRPGWKP RHGIPNNQKD TVCEDMTFST WTPPPGVHSQ TLSRFFDKVQ DLGRDSKTSS AEVTIQNVIK LVDELMEAPG DVEALAPPVR HLIATQLLSN LEDIMRILAK SLPKGPFTYI SPSNTELTLM IQERGDKNVT MGQSSARMKL NWAVAAGAED PGPAVAGILS IQNMTTLLAN ASLNLHSKKQ AELEEIYESS IRGVQLRRLS
CD97 is a G-protein-coupled seven-span transmembrane adhesive receptor that is constitutively expressed on granulocytes and monocytes and rapidly upregulated on T and B cells upon activation. CD97 is produced in alternatively spliced forms and its cellular ligand is CD55 (DAF), which protects various cell types from complement-mediated damage. Interaction of CD97 on leukocytes and CD55 on vessel cells probably facilitate leukocyte activation and migration into the tissues, similarly, CD97 seems to play a role in tumour migration and invasiveness. CD97 is involved in T cell regulation and peripheral granulocyte homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor E5; CD97; CD97 antigen; CD97 molecule; EGF-TM7 receptor; Leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; unnamed protein product
Gene Aliases: AA409984; ADGRE5; CD97; TM7LN1
UniProt ID: (Human) P48960, (Mouse) Q9Z0M6
Entrez Gene ID: (Human) 976, (Mouse) 26364
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support