Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CDO Antibodies

          Abnova

          CDON Polyclonal Antibody, MaxPab™

          View all (10) CDO antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CDON Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CDON Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CDON Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CDON Antibody (H00050937-B01P) in WB

          Western Blot analysis of CDON expression in transfected 293T cell line by CDON MaxPab polyclonal antibody. Lane 1: CDON transfected lysate(141.57 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CDON Antibody in Western Blot (WB)

          Product Details

          H00050937-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CDON (AAH98583.1, 1 a.a. approximately 1287 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MHPDLGPLCT LLYVTLTILC SSVSSDLAPY FTSEPLSAVQ KLGGPVVLHC SAQPVTTRIS WLHNGKTLDG NLEHIKIHQG TLTILSLNSS LLGYYQCLAN NSIGAIVSGP ATVSVAVLGD FGSSTKHVIT AEEKSAGFIG CRVPESNPKA EVRYKIRGKW LEHSTENYLI LPSGNLQILN VSLEDKGSYK CAAYNPVTHQ LKVEPIGRKL LVSRPSSDDV HILHPTHSQA LAVLSRSPVT LECVVSGVPA PQVYWLKDGQ DIAPESNWRR LYSHLATDSV DPADSGNYSC MAGNKSGDVK YVTYMVNVLE HASISKGLQD QIVSLGATVH FTCDVHGNPA PNCTWFHNAQ PIHPSARHLT AGNGLKISGV TVEDVGMYQC VADNGIGFMH STGRLEIEND GGFKPVIITA PVSAKVADGD FVTLSCNASG LPVPVIRWYD SHGLITSHPS QVLRSKSRKS QLSRPEGLNL EPVYFVLSQA GASSLHIQAV TQEHAGKYIC EAANEHGTTQ AEASLMVVPF ETNTKAETVT LPDAAQNDDR SKRDGSETGL LSSFPVKVHP SAVESAPEKN ASGISVPDAP IILSPPQTHT PDTYNLVWRA GKDGGLPINA YFVKYRKLDD GVGMLGSWHT VRVPGSENEL HLAELEPSSL YEVLMVARSA AGEGQPAMLT FRTSKEKTAS SKNTQASSPP VGIPKYPVVS EAANNNFGVV LTDSSRHSGV PEAPDRPTIS TASETSVYVT WIPRANGGSP ITAFKVEYKR MRTSNWLVAA EDIPPSKLSV EVRSLEPGST YKFRVIAINH YGESFRSSAS RPYQVVGFPN RFSSRPITGP HIAYTEAVSD TQIMLKWTYI PSSNNNTPIQ GFYIYYRPTD SDNDSDYKRD VVEGSKQWHM IGHLQPETSY DIKMQCFNEG GESEFSNVMI CETKVKRVPG ASEYPVKDLS TPPNSLGSGG NVGPATSPAR SSDMLYLIVG CVLGVMVLIL MVFIAMCLWK NRQQNTIQKY DPPGYLYQGS DMNGQMVDYT TLSGASQING NVHGGFLTNG GLSSGYSHLH HKVPNAVNGI VNGSLNGGLY SGHSNSLTRT HVDFEHPHHL VNGGGMYTAV PQIDPLECVN CRNCRNNNRC FTKTNSTFSS SPPPVVPVVA PYPQDGLEMK PLSHVKVPVC LTSAVPDCGQ LPEESVKDNV EPVPTQRTCC QDIVNDVSSD GSEDPAEFSR GQEGMINLRI PDHLQLAKSC VWEGDSCAHS ETENNIVSWN ALILPPVPEG CAEKTMWSPP GIPLDSPTEV LQQPRET

          Target Information

          CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cdon homolog; Cell adhesion molecule-related/down-regulated by oncogenes; immunoglobulin superfamily member; surface glycoprotein, Ig superfamily member

          View more View less

          Gene Aliases: CDO; CDON; CDON1; HPE11; Ihog; ORCAM

          View more View less

          UniProt ID: (Human) Q4KMG0

          View more View less

          Entrez Gene ID: (Human) 50937

          View more View less

          Function(s)
          protein binding coreceptor activity
          Process(es)
          cell fate specification positive regulation of neuroblast proliferation lens development in camera-type eye cell adhesion smoothened signaling pathway nervous system development neuroblast proliferation skeletal muscle tissue development myoblast fusion anterior/posterior pattern specification embryonic body morphogenesis skeletal muscle satellite cell differentiation central nervous system neuron differentiation cerebral cortex development neuron differentiation positive regulation of MAPK cascade regulation of neuron differentiation positive regulation of neuron differentiation positive regulation of transcription from RNA polymerase II promoter embryonic morphogenesis positive regulation of skeletal muscle tissue development positive regulation of small GTPase mediated signal transduction striated muscle cell differentiation embryonic retina morphogenesis in camera-type eye neural precursor cell proliferation negative regulation of biomineral tissue development cellular response to vitamin D negative regulation of canonical Wnt signaling pathway cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline