Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • C/EBP zeta Antibodies

          Abnova

          CEBPZ Polyclonal Antibody, MaxPab™

          View all (13) C/EBP zeta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CEBPZ Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CEBPZ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CEBPZ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CEBPZ Antibody (H00010153-B01P) in WB

          CEBPZ MaxPab polyclonal antibody. Western Blot analysis of CEBPZ expression in PC-12. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CEBPZ Antibody in Western Blot (WB)
          CEBPZ Polyclonal Antibody, MaxPab™

          Product Details

          H00010153-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CEBPZ (NP_005751.2, 1 a.a. approximately 1054 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAVKEPLEF HAKRPWRPEE AVEDPDEEDE DNTSEAENGF SLEEVLRLGG TKQDYLMLAT LDENEEVIDG GKKGAIDDLQ QGELEAFIQN LNLAKYTKAS LVEEDEPAEK ENSSKKEVKI PKINNKNTAE SQRTSVNKVK NKNRPEPHSD ENGSTTPKVK KDKQNIFEFF ERQTLLLRPG GKWYDLEYSN EYSLKPQPQD VVSKYKTLAQ KLYQHEINLF KSKTNSQKGA SSTWMKAIVS SGTLGDRMAA MILLIQDDAV HTLQFVETLV NLVKKKGSKQ QCLMALDTFK ELLITDLLPD NRKLRIFSQR PFDKLEQLSS GNKDSRDRRL ILWYFEHQLK HLVAEFVQVL ETLSHDTLVT TKTRALTVAH ELLCNKPEEE KALLVQVVNK LGDPQNRIAT KASHLLETLL CKHPNMKGVV SGEVERLLFR SNISSKAQYY AICFLNQMAL SHEESELANK LITVYFCFFR TCVKKKDVES KMLSALLTGV NRAYPYSQTG DDKVREQIDT LFKVLHIVNF NTSVQALMLL FQVMNSQQTI SDRYYTALYR KMLDPGLMTC SKQAMFLNLV YKSLKADIVL RRVKAFVKRL LQVTCQQMPP FICGALYLVS EILKAKPGLR SQLDDHPESD DEENFIDAND DEDMEKFTDA DKETEIVKKL ETEETVPETD VETKKPEVAS WVHFDNLKGG KQLNKYDPFS RNPLFCGAEN TSLWELKKLS VHFHPSVALF AKTILQGNYI QYSGDPLQDF TLMRFLDRFV YRNPKPHKGK ENTDSVVMQP KRKHFIKDIR HLPVNSKEFL AKEESQIPVD EVFFHRYYKK VAVKEKQKRD ADEESIEDVD DEEFEELIDT FEDDNCFSSG KDDMDFAGNV KKRTKGAKDN TLDEDSEGSD DELGNLDDDE VSLGSMDDEE FAEVDEDGGT FMDVLDDESE SVPELEVHSK VSTKKSKRKG TDDFDFAGSF QGPRKKKRNL NDSSLFVSAE EFGHLLDENM GSKFDNIGMN AMANKDNASL KQLRWEAERD DWLHNRDAKS IIKKKKHFKK KRIKTTQKTK KQRK

          Target Information

          This gene belongs to the CEBP family. The encoded protein plays a role in cellular response to environmental stimuli through a transcriptional process that involves heat shock factors, conserved DNA elements (heat shock elements or HSEs) and CCAAT boxes. The protein acts as a DNA-binding transcriptional activator and regulates the heat-shock protein 70 (HSP70) promoter in a CCAAT-dependent manner. The protein is also involved in cell growth and differentiation, particularly, hematopoietic differentiation. Methylation of the promoter of this gene or mutations within the gene may be correlated with occurrence of acute myeloid leukemia (AML).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C EBP; C/EBP zeta; CBF; CCAAT-binding factor; CCAAT-binding transcription factor 2; CCAAT-box-binding transcription factor; CCAAT/enhancer binding protein (C/EBP), zeta; CCAAT/enhancer binding protein alpha (C/EBP) related sequence 1; CCAAT/enhancer binding protein cebp family zeta; CCAAT/enhancer-binding protein zeta; CEBP zeta; core-binding factor; CTF2; HSPCBF; unnamed protein product

          View more View less

          Gene Aliases: AI848081; CBF; CBF2; Cebpa-rs1; CEBPZ; HSP-CBF; NOC1

          View more View less

          UniProt ID: (Human) Q03701, (Mouse) P53569

          View more View less

          Entrez Gene ID: (Human) 10153, (Mouse) 12607, (Rat) 362686

          View more View less

          Function(s)
          DNA binding transcription coactivator activity RNA binding protein binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding poly(A) RNA binding DNA-binding transcription factor gene-specific transcriptional regulator
          Process(es)
          positive regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline