Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: RENTDLREE IKVMERFRLD AWKRAEAIES SLEVEKKDKL ANTSVRFQNS QISSGTQVEK YKKNYLPMQG NPPRRSKSAP PRDLGNLDKG QAASPREPLE PLNFPDPEYK EEEEDQDIQG EISHPDGKVE KVYKNGCRVI LFPNGTRKEV SADGKTITVT FFNGDVKQVM PDQRVIYYYA AAQTTHTTYP EGLEVLHFSS GQIEKHYPDG RKEITFPDQT VKNLFPDGQE ESIFPDGTIV RVQRDGNKLI EFNNGQRELH TAQFKRREYP DGTVKTVYAN GHQETKYRSG RIRVKDKEGN VLMDTEL (1033-1338 aa encoded by BC024209)
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintenance of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-J; centromere protein J; LAG-3-associated protein; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7; microcephaly; Spindle assembly abnormal 4; unnamed protein product
Gene Aliases: 4932437H03Rik; BM032; CENP-J; CENPJ; Gm81; LAP; LIP1; MCPH6; Sas-4; Sas4; SASS4; SCKL4
UniProt ID: (Human) Q9HC77, (Mouse) Q569L8
Entrez Gene ID: (Human) 55835, (Rat) 305909, (Mouse) 219103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support