Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPTSSSFKH RIKEQEDYIR DWTAHREEIA RISQDLALIA REINDVAGEI DSVTSSGTAP STTLVDRVFD ESLNFQKIPP LVHSKTPEGN NGRSGDPRPQ AAEPPDHLTI TRRRTWSRDE VMGDNLLLSS VFQFSKKIRQ SIDKTAGKIR ILFKDKDRNW DDIESKLRAE SEVPIVKTSS MEISSILQEL KRVEKQLQAI NAMIDPDGTL EALNNMGFPS AMLPSPPKQK SSPVNNHHSP GQTPTLGQPE ARALHPAAVS AAAEFENAES EADFSIHFNR VNPDGEEEDV TVHK (1-293 aa encoded by BC014590)
This locus appears to be a transcribed pseudogene similar to centrosomal protein 170 kDa (CEP170). An approximately 50 kb region upstream of this locus also is homologous to CEP170, but is not transcribed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEP170 pseudogene 1; Cep170-like protein
Gene Aliases: CEP170L; FAM68B; KIAA0470L
Entrez Gene ID: (Human) 645455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support