Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: RKLKNSQEA QQRQATLVRK LQAKVLQYRS WCQELEKRLE ATGGPIPQRW ENVEEPNLDE LLVRLEEEQQ RCESLAEVNT QLRLHMEKAD VVNKALREDV EKLTVDWSRA RDELMRKESQ WQMEQEFFKG YLKGEHGRLL SLWREVVTFR RHFLEMKSAT DRDLMELKAE HVRLSGSLLT CCLRLTVGAQ SREPNGSGRM DGREPAQLLL LLAKTQELEK EAHERSQELI QLKSQGDLEK AELQDRVTEL SALLT (40-293 aa encoded by B C001433)
Probably plays an important role in centrosome cohesion during interphase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 250 kDa centrosomal protein; centrosomal Nek2-associated protein 1; unnamed protein product
Gene Aliases: C-NAP1; CEP2; CNAP1; CRDHL2
UniProt ID: (Human) Q9BV73
Entrez Gene ID: (Human) 11190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support