    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • MOB3B Antibodies
          OriGene
          MOBKL2B Monoclonal Antibody (OTI1G1), TrueMAB™
          View all (2) MOB3B antibodies
          Tech Support
          Cite MOBKL2B Monoclonal Antibody (OTI1G1), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          MOBKL2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MOBKL2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          MOBKL2B Antibody (CF501499) in ICC/IF
          Anti-MOBKL2B mouse monoclonal antibody (TA501499) Immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY MOBKL2B(RC205977). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MOBKL2B Antibody in Immunocytochemistry (ICC/IF)
          MOBKL2B Antibody in Western Blot (WB)
          MOBKL2B Antibody in Flow Cytometry (Flow)
          MOBKL2B Monoclonal Antibody (OTI1G1), TrueMAB™
          Product Details
          CF501499
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI1G1
          Immunogen
          Full length human recombinant protein of human MOBKL2B produced in HEK293T cell.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 8% trehalose
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
          Target Information
          The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.
          Bioinformatics
          Protein Aliases: MOB kinase activator 3B; MOB kinase activator-like 2B; Mob1 homolog 2b; MOB1, Mps One Binder kinase activator-like 2B; MOBKL2B; monopolar spindle 1 binding, MOB1, domain containing; Mps one binder kinase activator-like 2B; unnamed protein product
          View more View less
          Gene Aliases: C9orf35; MOB1D; MOB3B; MOBKL2B
          View more View less
          UniProt ID: (Human) Q86TA1
          View more View less
          Entrez Gene ID: (Human) 79817
          View more View less
          Function(s)
          protein binding protein kinase activator activity metal ion binding
          Process(es)
          signal transduction regulation of hippo signaling
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline