FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
ZNF500 is a new candidate transcription factor. It is derived from an annotated genomic sequence (NT_037887) using a gene prediction method.
Protein Aliases: unnamed protein product; Zinc finger protein 500; Zinc finger protein with KRAB and SCAN domains 18
Gene Aliases: KIAA0557; ZKSCAN18; ZNF500; ZSCAN50
UniProt ID: (Human) O60304
Entrez Gene ID: (Human) 26048