Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLLLINVIL TLWVSCANGQ VKPCDFPDIK HGGLFHENMR RPYFPVAVGK YYSYYCDEHF ETPSGSYWDY IHCTQNGWSP AVPCLRKCYF PYLENGYNQN YGRKFVQGNS TEVACHPGYG LPKAQTTVTC TEKGWSPTPR CIRVRTCSKS DIEIENGFIS ESSSIYILNK EIQYKCKPGY ATADGNSSGS ITCLQNGWSA QPICINSSEK CGPPPPISNG DTTSFLLKVY VPQSRVEYQC QPYYELQGSN YVTCSNGEWS EPPRCIHPCI ITEENMNKNN IKLKGRSDRK YYAKTGDTIE FMCKLGYNAN TSILSFQAVC REGIVEYPRC E (1-330 aa encoded by BC058009)
The protein encoded by this gene is a secreted protein, which belongs to the complement factor H-related protein family. It binds to heparin, and may be involved in complement regulation. Mutations in this gene are associated with decreased risk of age-related macular degeneration, and with an increased risk of atypical hemolytic-uremic syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complement factor H-related protein 3; DOWN16; FHR-3; H factor-like 4; H factor-like protein 3; unnamed protein product
Gene Aliases: CFHL3; CFHR3; DOWN16; EG624286; FHR-3; FHR3; HLF4
UniProt ID: (Human) Q02985
Entrez Gene ID: (Human) 10878, (Mouse) 624286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support