Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MKQESAAPN TPPTSQSPTP SAQFPRNDGD PQALWIFGYG SLVWRPDFAY SDSRVGFVRG YSRRFWQGDT FHRGSDKMPG RVVTLLEDHE GCTWGVAYQV QGEQVSKALK YLNVREAVLG GYDTKEVTFY PQDAPDQPLK ALAYVATPQN PGYLGPAPEE AIATQILACR GFSGHNLEYL LRLADFMQLC GPQAQDEHLA AIVDAVGTML PCFCPTEQAL ALV (1-222 aa encoded by BC001847)
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: blocks Notch protein; botch; cation transport regulator homolog 1; Cation transport regulator-like protein 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator homolog 1 (E. coli); ChaC, cation transport regulator-like 1; gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; hchaC-LP; neuroprotective protein 7; NPG7; unnamed protein product
Gene Aliases: 1810008K03Rik; RGD1307153
UniProt ID: (Human) Q9BUX1, (Rat) B3STU3, (Mouse) Q8R3J5
Entrez Gene ID: (Human) 79094, (Chinese hamster) 100762174, (Rat) 362196, (Mouse) 69065
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support