Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MGGTTSTRR VTFEADENEN ITVVKGIRLS ENVIDRMKES SPSGSKSQRY SGAYGASVSD EELKRRVAEE LALEQAKKES EDQKRLKQAK ELDRERAAAN EQLTRAILRE RICSEEERAK AKHLARQLEE KDRVLKKQDA FYKEQLARLE ERSSEFYRVT TEQYQKAAEE VEAKFKRYES HPVCADLQAK ILQCYRENTH QTLKCSALAT QYMHCVNHAK QSMLEKGG (1-227 aa encoded by BC014839)
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; mitochondrial contact site and cristae organizing system subunit 19; mitochondrial inner membrane organizing system 3; protein phosphatase 1, regulatory subunit 22; unnamed protein product
Gene Aliases: 0610041L09Rik; 1700039J09Rik; Mic19; MICOS19; MINOS3; PPP1R22
UniProt ID: (Human) Q9NX63, (Mouse) Q9CRB9
Entrez Gene ID: (Human) 54927, (Mouse) 66075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support