Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CHD7 Antibodies

          Bethyl Laboratories

          CHD7 Polyclonal Antibody

          View all (9) CHD7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CHD7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CHD7 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          CHD7 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CHD7 Antibody (A301-222A) in IP

          Detection of human CHD7 by western blot of immunoprecipitates. Samples: Whole cell lysate (1.0 mg per IP reaction; 20% of IP loaded) from HEK293T cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-CHD7 antibody (Product # A301-222A) (lot 2) used for IP at 3 µg per reaction. CHD7 was also immunoprecipitated by rabbit anti-CHD7 antibody A301-223A. For blotting immunoprecipitated CHD7, A301-223A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 75 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CHD7 Antibody in Immunoprecipitation (IP)
          CHD7 Polyclonal Antibody

          Product Details

          A301-222A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          2-10 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 2025 and 2075 of human chromodomain helicase DNA binding protein 7.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris citrate/phosphate, pH 7-8

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Target Information

          Chromodomain helicase DNA binding protein 7 (CHD7) is a gene located on chromosome 8q12.2 that encodes a member of the chromodomain helicase DNA-binding (CHD) family, which plays a pivotal role in chromatin remodeling and gene expression regulation. CHD7 contains multiple domains that facilitate its involvement in modifying chromatin structure, thereby influencing the accessibility of DNA for transcription factors and other regulatory proteins. The protein is integral to various developmental processes, including neural crest formation, thus affecting tissues such as the nervous system, skeletal structures, and sensory organs. Mutations in CHD7 are associated with CHARGE syndrome, a multisystem disorder characterized by congenital anomalies including coloboma, heart defects, and ear abnormalities. CHD7's role in chromatin dynamics makes it crucial for normal embryonic development and cellular differentiation. Ongoing research is focused on uncovering CHD7's broader implications in developmental biology and its potential as a therapeutic target in chromatin-related disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP-dependent chromatin remodeler CHD7; ATP-dependent helicase CHD7; CHARGE association; CHD-7; Chromo domain-containing protein 7; unnamed protein product

          View more View less

          Gene Aliases: CHD7; CRG; HH5; IS3; KAL5; KIAA1416

          View more View less

          UniProt ID: (Human) Q9P2D1

          View more View less

          Entrez Gene ID: (Human) 55636

          View more View less

          Function(s)
          nucleotide binding RNA polymerase II core promoter proximal region sequence-specific DNA binding DNA binding chromatin binding protein binding ATP binding hydrolase activity ATPase activity histone binding ATP-dependent chromatin remodeler activity promoter-specific chromatin binding
          Process(es)
          skeletal system development blood vessel development in utero embryonic development blood vessel remodeling heart morphogenesis ventricular trabecula myocardium morphogenesis right ventricular compact myocardium morphogenesis chromatin organization chromatin remodeling regulation of transcription, DNA-templated rRNA processing transcription from RNA polymerase II promoter central nervous system development adult heart development sensory perception of sound locomotory behavior adult walking behavior blood circulation response to bacterium regulation of gene expression regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum cranial nerve development olfactory nerve development olfactory bulb development T cell differentiation female genitalia development embryonic hindlimb morphogenesis aorta development aorta morphogenesis atrioventricular canal development positive regulation of multicellular organism growth olfactory behavior ear morphogenesis inner ear morphogenesis chordate embryonic development camera-type eye development nose development positive regulation of transcription from RNA polymerase II promoter semicircular canal morphogenesis tissue remodeling genitalia development artery morphogenesis regulation of neurogenesis neurological system process cognition regulation of transport palate development retina development in camera-type eye regulation of growth hormone secretion limb development head development face development innervation cardiac septum morphogenesis epithelium development secondary palate development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline