Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHD7 Antibodies

          Invitrogen

          CHD7 Polyclonal Antibody

          View all (10) CHD7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CHD7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CHD7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CHD7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CHD7 Antibody (PA5-47918) in ICC/IF

          Immunocytochemistry analysis of CHD7 in immersion fixed HepG2 human hepatocellular carcinoma cell line. Samples were incubated in CHD7 polyclonal antibody (Product # PA5-47918) using a dilution of 10 µg/mL for 3 hours at room temperature followed by NorthernLights™ 557-conjugated Anti-Sheep IgG Secondary Antibody (red, upper panel) and counterstained with DAPI (blue, lower panel). Specific staining was localized to nuclei. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CHD7 Antibody in Immunocytochemistry (ICC/IF)
          CHD7 Antibody in Western Blot (WB)
          CHD7 Polyclonal Antibody

          Product Details

          PA5-47918

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5-15 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Sheep / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E. coli-derived recombinant human CHD7 Ala263-Gln457
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2576825

          Product Specific Information

          Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

          Target Information

          Chromodomain helicase DNA binding protein 7 (CHD7) is a gene located on chromosome 8q12.2 that encodes a member of the chromodomain helicase DNA-binding (CHD) family, which plays a pivotal role in chromatin remodeling and gene expression regulation. CHD7 contains multiple domains that facilitate its involvement in modifying chromatin structure, thereby influencing the accessibility of DNA for transcription factors and other regulatory proteins. The protein is integral to various developmental processes, including neural crest formation, thus affecting tissues such as the nervous system, skeletal structures, and sensory organs. Mutations in CHD7 are associated with CHARGE syndrome, a multisystem disorder characterized by congenital anomalies including coloboma, heart defects, and ear abnormalities. CHD7's role in chromatin dynamics makes it crucial for normal embryonic development and cellular differentiation. Ongoing research is focused on uncovering CHD7's broader implications in developmental biology and its potential as a therapeutic target in chromatin-related disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP-dependent chromatin remodeler CHD7; ATP-dependent helicase CHD7; CHARGE association; CHD-7; Chromo domain-containing protein 7; unnamed protein product

          View more View less

          Gene Aliases: CHD7; CRG; HH5; IS3; KAL5; KIAA1416

          View more View less

          UniProt ID: (Human) Q9P2D1

          View more View less

          Entrez Gene ID: (Human) 55636

          View more View less

          Function(s)
          nucleotide binding RNA polymerase II core promoter proximal region sequence-specific DNA binding DNA binding chromatin binding protein binding ATP binding hydrolase activity ATPase activity histone binding ATP-dependent chromatin remodeler activity promoter-specific chromatin binding
          Process(es)
          skeletal system development blood vessel development in utero embryonic development blood vessel remodeling heart morphogenesis ventricular trabecula myocardium morphogenesis right ventricular compact myocardium morphogenesis chromatin organization chromatin remodeling regulation of transcription, DNA-templated rRNA processing transcription from RNA polymerase II promoter central nervous system development adult heart development sensory perception of sound locomotory behavior adult walking behavior blood circulation response to bacterium regulation of gene expression regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum cranial nerve development olfactory nerve development olfactory bulb development T cell differentiation female genitalia development embryonic hindlimb morphogenesis aorta development aorta morphogenesis atrioventricular canal development positive regulation of multicellular organism growth olfactory behavior ear morphogenesis inner ear morphogenesis chordate embryonic development camera-type eye development nose development positive regulation of transcription from RNA polymerase II promoter semicircular canal morphogenesis tissue remodeling genitalia development artery morphogenesis regulation of neurogenesis neurological system process cognition regulation of transport palate development retina development in camera-type eye regulation of growth hormone secretion limb development head development face development innervation cardiac septum morphogenesis epithelium development secondary palate development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline