Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CHKB Antibodies

          Abnova

          CHKB Polyclonal Antibody, MaxPab™

          View all (6) CHKB antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CHKB Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CHKB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CHKB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CHKB Antibody (H00001120-D01P) in WB

          Western Blot analysis of CHKB expression in transfected 293T cell line by CHKB MaxPab polyclonal antibody. Lane 1: CHKB transfected lysate(45.30 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CHKB Antibody in Western Blot (WB)
          CHKB Polyclonal Antibody, MaxPab™

          Product Details

          H00001120-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CHKB (NP_005189.2, 1 a.a. approximately 395 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAEATAVAG SGAVGGCLAK DGLQQSKCPD TTPKRRRASS LSRDAERRAY QWCREYLGGA WRRVQPEELR VYPVSGGLSN LLFRCSLPDH LPSVGEEPRE VLLRLYGAIL QGVDSLVLES VMFAILAERS LGPQLYGVFP EGRLEQYIPS RPLKTQELRE PVLSAAIATK MAQFHGMEMP FTKEPHWLFG TMERYLKQIQ DLPPTGLPEM NLLEMYSLKD EMGNLRKLLE STPSPVVFCH NDIQEGNILL LSEPENADSL MLVDFEYSSY NYRGFDIGNH FCEWVYDYTH EEWPFYKARP TDYPTQEQQL HFIRHYLAEA KKGETLSQEE QRKLEEDLLV EVSRYALASH FFWGLWSILQ ASMSTIEFGY LDYAQSRFQF YFQQKGQLTS VHSSS

          Target Information

          The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase, the initial enzyme in the sequence, plays a role in cell growth proliferation. A related protein, ChoKB (also, known as choline kinase beta), is a 395 amino acid enzyme that catalyzes the phosphorylation of choline by ATP in the presence of magnesium, thereby yielding phosphocholine and ADP. Like all choline kinases, ChoKB possesses ethanoalamine kinase activity and catalyzes the phosphorylation of ethanolamine. The gene encoding ChoKB is located less than 1 kb upstream of the CPT1B gene, suggesting that the ChoKB gene may regulate transcription CPT1B. In mice, mutations and/or deletions in the gene encoding ChoKB are the cause of hindlimb muscular dystrophy and neonatal bone deformity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CHKL; Choline kinase beta; Choline kinase-like protein; Choline/ethanolamine kinase; choline/ethanolamine kinase beta; CK; CKEKB; EK; EKB; Ethanolamine kinase; Ethanolamine kinase beta; unnamed protein product

          View more View less

          Gene Aliases: CHETK; CHKB; CHKL; CK; CKB; CKEKB; EK; EKB; MDCMC

          View more View less

          UniProt ID: (Human) Q9Y259

          View more View less

          Entrez Gene ID: (Human) 1120

          View more View less

          Function(s)
          nucleotide binding choline kinase activity ethanolamine kinase activity ATP binding kinase activity transferase activity
          Process(es)
          lipid metabolic process phosphatidylethanolamine biosynthetic process phosphatidylcholine biosynthetic process CDP-choline pathway muscle organ development phospholipid biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-w8jw2:80/100.66.76.154:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline