Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is highly specific to the CHMP4B and it doesn't cross-react with other CHMPs as well as other CHMP4s (CHMP4A and CHMP4C).
Immunogen sequence: MSVFGKLFG AGGGKAGKGG PTPQEAIQRL RDTEEMLSKK QEFLEKKIEQ ELTAAKKHGT KNKRAALQAL KRKKRYEKQL AQIDGTLSTI EFQREALENA NTNTEVLKNM GYAAKAMKAA HDNMDIDKVD ELMQDIADQQ ELAEEISTAI SKPVGFGEEF DEDELMAELE ELEQEELDKN LLEISGPETV PLPNVPSIAL PSKPAKKKEE EDDDMKELEN WAGSM (1-224 aa encoded by BC033859)
CHMP4B is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) protein family. The protein is part of the endosomal sorting complex required for transport (ESCRT) complex III (ESCRT-III), which functions in the sorting of endocytosed cell-surface receptors into multivesicular endosomes. The ESCRT machinery also functions in the final abscisson stage of cytokinesis and in the budding of enveloped viruses such as HIV-1. The three proteins of the CHMP4 subfamily interact with programmed cell death 6 interacting protein (PDCD6IP, also known as ALIX), which also functions in the ESCRT pathway. The CHMP4 proteins assemble into membrane-attached 5-nm filaments that form circular scaffolds and promote or stabilize outward budding. These polymers are proposed to help generate the luminal vesicles of multivesicular bodies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromatin modifying protein 4B; chromatin-modifying protein 4b; similar to yeast Snf7p; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; unnamed protein product; vacuolar protein-sorting-associated protein 32-2
Gene Aliases: 2010012F05Rik; C20orf178; CHMP4A; CTPP3; CTRCT31; dJ553F4.4; RGD1309846; RGD1565889; Shax1; SNF7; SNF7-2; Vps32-2; VPS32B
UniProt ID: (Human) Q9H444, (Mouse) Q9D8B3
Entrez Gene ID: (Human) 128866, (Mouse) 75608, (Rat) 100359642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support