Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
PA1-32702 detects CHRM2 from human samples.
Muscarinic cholinergic receptor 2 (CHRM2) is an Acetylcholine Receptor has been suggested to function as a presynaptic autoreceptor that inhibits acetylcholine release. The M2 receptor has also been suggested to mediate smooth muscle contractility. Alzheimer's disease (AD) involves selective loss of M2 subtype receptors in cortical and hippocampal regions of the human brain. Increase in M2 receptor heterozygosity has been associated with depression in women. The M2 receptor has been reported to be expressed in various regions of the brain including the cerebellum, cerebral cortex, hippocampus, medulla, striatum, and thalamus, and in lung and prostate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7TM receptor; acetylcholine receptor; acetylcholine receptor, muscarinic 2; AChR M2; cardiac muscarinic acetylcholine receptor; cholinergic receptor; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, cardiac; cytotransmitter gated ion channel; FLJ43243; G protein coupled receptor; M2 muscarinic acetylcholine receptor; M2-mAChR; m2AChR; MGC120006; MGC120007; muscarinic acetylcholine receptor 2; Muscarinic acetylcholine receptor M2; muscarinic acetylcholine receptor M2-like; muscarinic cholinergic receptor; muscarinic M2 receptor; muscarinic receptor 2; muscarinic receptor m2; unnamed protein product
Gene Aliases: AChR-M2; Acm2; Chrm-2; Gpm2; HM2; I79_007306; M2
UniProt ID: (Human) P08172, (Bovine) P41985, (Pig) P06199, (Rat) P10980, (Mouse) Q9ERZ4
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support