Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody (10209-2-AP) is a rabbit polyclonal antibody raised against the full-length CIRBP of human origin.
Immunogen sequence: MASDEGKLF VGGLSFDTNE QSLEQVFSKY GQISEVVVVK DRETQRSRGF GFVTFENIDD AKDAMMAMNG KSVDGRQIRV DQAGKSSDNR SRGYRGGSAG GRGFFRGGRG RGRGFSRGGG DRGYGGNRFE SRSGGYGGSR DYYSSRSQSG GYSDRSSGGS YRDSYDSYAT HNE (1-172 aa encoded by BC000403)
CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A18 hnRNP; CIRP; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; testicular tissue protein Li 39; unnamed protein product
Gene Aliases: CIRP
UniProt ID: (Human) Q14011, (Mouse) P60824, (Rat) P60825
Entrez Gene ID: (Human) 1153, (Mouse) 12696, (Rat) 81825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support