    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • CD27 Antibodies
          Proteintech
          CD27 Monoclonal Antibody (O323), CoraLite® Plus 405
          View all (200) CD27 antibodies
          Tech Support
          Cite CD27 Monoclonal Antibody (O323), CoraLite® Plus 405
          • Antibody Testing Data (2)
          CD27 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD27 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD27 Antibody (CL405-65132) in Flow
          1X10^6 human PBMCs were surface stained with 5 µL CoraLite® Plus 405 Anti-Human CD27 (CL405-65132, Clone:O323) (red), or Mouse IgG1 Isotype Control. Cells were not fixed. Lymphocytes were gated. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD27 Antibody in Flow Cytometry (Flow)
          CD27 Antibody in Flow Cytometry (Flow)
          Product Details
          CL405-65132
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          O323
          Immunogen
          The exact sequence is proprietary
          Conjugate
          CoraLite® Plus 405 CoraLite® Plus 405 CoraLite® Plus 405
          • APC 
          • Atlantic Blue
          • Cardinal Red
          • CoraLite Plus 488
          • CoraLite Plus 594
          • CoraLite Plus 647
          • CoraLite Plus 670
          • CoraLitePlus 555
          • FITC Plus
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          399/422 nm
          Form
          Liquid
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 0.5% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          CD27 is a 50 kDa member of the tumor necrosis factor (TNF) receptor superfamily that includes CD40 and CD30. The TNF superfamily members are known for the regulation of cell proliferation and death. In contrast to the expression of other TNFR/TNF family members, expression of CD27 and its ligand CD70 is predominantly confined to lymphocytes. High expression levels of CD27 appear to be dependent on proper ligation of antigen receptors. CD70 expression requires additional co-stimulatory and/or pro-inflammatory signals. CD27 is expressed as a disulfide-linked homodimer on mature thymocytes, peripheral blood T cells and a subpopulation of B cells. Activation of T cells via TCR-CD3 complex results in upregulation of CD27 expression on the plasma membrane as well as in the release of its soluble 28-32 kDa form, sCD27, detected in the plasma, urine or spinal fluid. Soluble CD27 is an important prognostic marker of acute and chronic B cell malignancies. RgpA, a cystein proteinase, although activating T cells through the protease-activated receptors (PARs), degradates CD27 and counteracts T cell activation mediated by CD27 and its ligand CD70. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor. Diseases associated with CD27 dysfunction include Lymphoproliferative Syndrome 2 and Autosomal Recessive Lymphoproliferative Syndrome.
          Bioinformatics
          Protein Aliases: CD27; CD27 antigen; CD27L receptor; LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T-cell activation antigen CD27; T14; TNFSF7; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7
          View more View less
          Gene Aliases: CD27; S152; S152. LPFS2; T14; TNFRSF7; Tp55
          View more View less
          UniProt ID: (Human) P26842
          View more View less
          Entrez Gene ID: (Human) 939
          View more View less
          Function(s)
          transmembrane signaling receptor activity tumor necrosis factor-activated receptor activity protein binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          Process(es)
          inflammatory response immune response cell surface receptor signaling pathway brain development release of cytoplasmic sequestered NF-kappaB extrinsic apoptotic signaling pathway via death domain receptors immunoglobulin mediated immune response response to lipopolysaccharide tumor necrosis factor-mediated signaling pathway regulation of cell proliferation negative regulation of apoptotic process negative regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of MAPK cascade response to ethanol positive regulation of B cell differentiation positive regulation of JNK cascade negative regulation of T cell apoptotic process extrinsic apoptotic signaling pathway
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-ngnjr:80/100.66.79.104:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0