Immunogen sequence: RAAYLFIGP SIDVQSFTVH GDSQQLDVFI ATSKHAYATY LWTGEATGQS AFAQVIADRH KILFDRNSAI KSSIVPEVKD YAAIVEQNLQ HFKPVFQLLE KQILSRVRNT ASFRKTAERL LRFSDKRQTE EAIDRIFAIS QQQQQQSKSK KNRRAGKRYK FVDAVPDIFA QIEVNEKKIR QKAQILAQKE LPIDEDEEMK DLLDFADVTY EKHKNGGLIK GRFGQARMVT TTHSRAPSLS ASYTRLFLIL NIAIFFVMLA MQLTYFQRAQ SLHGQRCLYA VLLIDSCILL WLYSSCSQSQ C
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
Protein Aliases: Chondroitin-glucuronate 5-epimerase; Dermatan-sulfate epimerase; DS epimerase; SART-2; Squamous cell carcinoma antigen recognized by T-cells 2; unnamed protein product
Gene Aliases: DS-epi1; DSE; DSEP; DSEPI; EDSMC2; SART-2; SART2
UniProt ID: (Human) Q9UL01
Entrez Gene ID: (Human) 29940