    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRIP6 Antibodies
          Proteintech
          TRIP6 Monoclonal Antibody (5D8C7), CoraLite® Plus 488
          View all (18) TRIP6 antibodies
          Tech Support
          Cite TRIP6 Monoclonal Antibody (5D8C7), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          TRIP6 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TRIP6 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TRIP6 Antibody (CL488-60205) in Flow
          1x10^6 HepG2 cells were intracellularly stained with 0.8 µg Coralite® Plus 488 Trip6 Monoclonal Antibody (CL488-60205, Clone: 5D8C7) (red), or 0.8 µg CoraLite® Plus 488 Mouse IgG1 Isotype Control (MOPC-21) (CL488-65124, Clone: MOPC-21) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRIP6 Antibody in Flow Cytometry (Flow)
          TRIP6 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-60205
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5D8C7
          Immunogen
          TRIP6 fusion protein Ag15149
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PPPAYRTGS LKPNPASPLP ASPYGGPTPA SYTTASTPAG PAFPVQVKVA QPVRGCGPPR RGASQASGPL PGPHFPLPGR GEVWGPGYRS QREPGPGAKE EAAGVSGPAG RGRGGEHGPQ VPLSQPPEDE LDRLTKKLVH DMNHPPSGEY FGQCGGCGED VVGDGAGVVA FDRVFHVGCF VCSTCRAQLR GQHFYAVERR AYCEGCYVAT LEKCATCSQP ILDRILRAMG KAYHPGCFTC VVCHRGLDGI PFTVDATSQI HCIEDFHRKF APRCSVCGGA IMPEPGQEET VRIVALDRSF HIGCYKCEEC GLLLSSEGEC QGCYPLDGHI LCKACSAWRI QELSATVTTD C
          Target Information
          This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
          Bioinformatics
          Protein Aliases: MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP-1; OPA-interacting protein 1; similar to ZRP-1 and TRIP6; tcag7.1300; thyroid hormone receptor interacting protein 6; Thyroid receptor-interacting protein 6; TR-interacting protein 6; TRIP-6; unnamed protein product; ZRP-1; zyxin related protein 1; Zyxin-related protein 1
          View more View less
          Gene Aliases: OIP-1; OIP1; TRIP-6; TRIP6; TRIP6i2; ZRP-1; Zrp1
          View more View less
          UniProt ID: (Human) Q15654, (Mouse) Q9Z1Y4
          View more View less
          Entrez Gene ID: (Human) 7205, (Mouse) 22051
          View more View less
          Function(s)
          RNA binding interleukin-1 receptor binding protein binding kinase binding metal ion binding thyroid hormone receptor binding zinc ion binding poly(A) RNA binding actin or actin-binding cytoskeletal protein
          Process(es)
          cell adhesion signal transduction positive regulation of cell migration chordate embryonic development focal adhesion assembly positive regulation of NIK/NF-kappaB signaling transcription, DNA-templated regulation of transcription, DNA-templated release of cytoplasmic sequestered NF-kappaB
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline