    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • FUT9 Antibodies
          Proteintech
          FUT9 Monoclonal Antibody (5D4G6), CoraLite® Plus 488
          Tech Support
          Cite FUT9 Monoclonal Antibody (5D4G6), CoraLite® Plus 488
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          FUT9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FUT9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FUT9 Antibody (CL488-60230) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed MCF-7 cells using CoraLite® Plus 488 FUT9 antibody (CL488-60230, Clone: 5D4G6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FUT9 Antibody in Immunocytochemistry (ICC/IF)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-60230
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5D4G6
          Immunogen
          FUT9 fusion protein Ag8298
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PTNSWIFSP MESASSVLKM KNFFSTKTDY FNETTILVWV WPFGQTFDLT SCQAMFNIQG CHLTTDRSLY NKSHAVLIHH RDISWDLTNL PQQARPPFQK WIWMNLESPT HTPQKSGIEH LFNLTLTYRR DSDIQVPYGF LTVSTNPFVF EVPSKEKLVC WVVSNWNPEH ARVKYYNELS KSIEIHTYGQ AFGEYVNDKN LIPTISACKF YLSFENSIHK DYITEKLYNA FLAGSVPVVL GPSRENYENY IPADSFIHVE DYNSPSELAK YLKEVDKNNK LYLSYFNWRK DFTVNLPRFW ESHACLACDH VKRHQEYKSV GNLEKWFWN
          Target Information
          Transfers a fucose to lacto-N-neotetraose but not to either alpha2,3-sialyl lacto-N-neotetraose or lacto-N-tetraose. Can catalyze the last step in the biosynthesis of Lewis antigen, the addition of a fucose to precursor polysaccharides.
          Bioinformatics
          Protein Aliases: alpha-(1,3)-fucosyltransferase 9; alpha-1,3-L-fucosyltransferase; fuc-TIX; fucosyltransferase 9 (alpha (1,3) fucosyltransferase); fucosyltransferase IX; fucT-IX; galactoside 3-L-fucosyltransferase; unnamed protein product
          View more View less
          Gene Aliases: Fuc-TIX; mFuc-TIX; mFUT9
          View more View less
          UniProt ID: (Human) Q9Y231, (Mouse) O88819, (Rat) Q99JB3
          View more View less
          Entrez Gene ID: (Pig) 102160171, (Human) 10690, (Mouse) 14348, (Rat) 84597
          View more View less
          Function(s)
          protein binding fucosyltransferase activity 4-galactosyl-N-acetylglucosaminide 3-alpha-L-fucosyltransferase activity protein homodimerization activity alpha-(1->3)-fucosyltransferase activity glycosyltransferase
          Process(es)
          polysaccharide biosynthetic process carbohydrate metabolic process protein N-linked glycosylation N-glycan processing protein O-linked glycosylation sphingolipid metabolic process glycosphingolipid biosynthetic process glycoprotein biosynthetic process oligosaccharide biosynthetic process positive regulation of neuron projection development neuron differentiation neuronal stem cell division L-fucose catabolic process Lewis x epitope biosynthetic process regulation of leukocyte cell-cell adhesion regulation of leukocyte tethering or rolling nervous system development
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0