    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PGRMC2 Antibodies
          Proteintech
          PGRMC2 Monoclonal Antibody (5F10D7), CoraLite® Plus 488
          View all (13) PGRMC2 antibodies
          Tech Support
          Cite PGRMC2 Monoclonal Antibody (5F10D7), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PGRMC2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          PGRMC2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PGRMC2 Antibody (CL488-60249) in Flow
          1x10^6 HepG2 cells were intracellularly stained with 0.4 µg CoraLite® Plus 488 Anti-Human PGRMC2 (CL488-60249, Clone: 5F10D7) (red), or 0.4 µg CoraLite® Plus 488 Mouse IgG2a Isotype Control (C1.18.4) (CL488-65208, Clone: C1.18.4) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PGRMC2 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-60249
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5F10D7
          Immunogen
          PGRMC2 fusion protein Ag20077
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite 594
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LLNVALVAL VLLGAYRLWV RWGRRGLGAG AGAGEESPAT SLPRMKKRDF SLEQLRQYDG SRNPRILLAV NGKVFDVTKG SKFYGPAGPY GI
          Target Information
          PGRMC2 (progesterone receptor membrane component 2), also known as DG6 (steroid receptor protein DG6) or PMBP (progesterone membrane-binding protein), is a single pass membrane protein belonging to the cytochrome b5 family (MAPR (membrane associated progesterone receptor) subfamily). Expressed in sperm, PGRMC2 is believed to function as a steroid receptor and may participate in the progesterone-dependent sperm acrosome reaction. PGRMC2 shares approximately 50% overall sequence identity with its close relative PGRMC1. The loss of the gene encoding PGRMC2 is associated with metastasis in uterine endocervical adenocarcinomas, implicating a potential role of PGRMC2 in the suppression of metastasis of endocervical adenocarcinomas.
          Bioinformatics
          Protein Aliases: Membrane-associated progesterone receptor component 2; progesterone membrane binding protein; Progesterone membrane-binding protein; Steroid receptor protein DG6; unnamed protein product
          View more View less
          Gene Aliases: 4631434O19Rik; 5730409G06Rik; DG6; PGRMC2; PMBP
          View more View less
          UniProt ID: (Human) O15173, (Mouse) Q80UU9
          View more View less
          Entrez Gene ID: (Human) 10424, (Mouse) 70804
          View more View less
          Function(s)
          steroid hormone receptor activity steroid binding protein binding lipid binding heme transporter activity heme binding transmembrane signal receptor
          Process(es)
          heme transport intracellular steroid hormone receptor signaling pathway adipose tissue development biological_process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0