    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PGM1 Antibodies
          Proteintech
          PGM1 Monoclonal Antibody (2G5A8), CoraLite® Plus 488
          View all (18) PGM1 antibodies
          Tech Support
          Cite PGM1 Monoclonal Antibody (2G5A8), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PGM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PGM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PGM1 Antibody (CL488-66105) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 PGM1 antibody (CL488-66105, Clone: 2G5A8) at dilution of 1:600. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PGM1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-66105
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:300-1:1,200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2G5A8
          Immunogen
          PGM1 fusion protein Ag7294
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: SGPNRLKIC IDAMHGVVGP YVKKILCEEL GAPANSAVNC VPLEDFGGHH PDPNLTYAAD LVETMKSGEH DFGAAFDGDG DRNMILGKHG FFVNPSDSVA VIAANIFSIP YFQQTGVRGF ARSMPTSGAL DRVASATKIA LYETPTGWKF FGNLMDASKL SLCGEESFGT GSDHIREKDG LWAVLAWLSI LATRKQSVED ILKDHWQKHG RNFFTRYDYE EVEAEGANKM MKDLEALMFD RSFVGKQFSA NDKVYTVEKA DNFEYSDPVD GSISRNQGLR LIFTDGSRIV FRLSGTGSAG ATIRLYIDSY EKDVAKINQD PQVMLAPLIS IALKVSQLQE RTGRTAPTVI T
          Target Information
          Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
          Bioinformatics
          Protein Aliases: Glucose phosphomutase 1; Glucose phosphomutase 2; PGM 1; PGM 2; PGM1; Phosphodeoxyribomutase; phosphoglucomutase 2; Phosphoglucomutase-1; Phosphoglucomutase-2; Phosphopentomutase; unnamed protein product
          View more View less
          Gene Aliases: 3230402E02Rik; CDG1T; GSD14; PGM; Pgm-1; PGM1; Pgm2
          View more View less
          UniProt ID: (Rabbit) P00949, (Human) P36871, (Mouse) Q7TSV4
          View more View less
          Entrez Gene ID: (Rabbit) 100009155, (Pig) 397566, (Human) 5236, (Rat) 24645, (Mouse) 66681
          View more View less
          Function(s)
          magnesium ion binding phosphoglucomutase activity protein binding isomerase activity intramolecular transferase activity, phosphotransferases metal ion binding molecular_function phosphopentomutase activity mutase
          Process(es)
          carbohydrate metabolic process glucose metabolic process gluconeogenesis glycolytic process galactose catabolic process via UDP-galactose glycogen biosynthetic process cellular calcium ion homeostasis response to radiation galactose catabolic process organic substance metabolic process
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline