    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Iduronate 2 Sulfatase Antibodies
          Proteintech
          Iduronate 2 sulfatase Monoclonal Antibody (1A3F9), CoraLite® Plus 488
          View all (24) Iduronate 2 Sulfatase antibodies
          Tech Support
          Cite Iduronate 2 sulfatase Monoclonal Antibody (1A3F9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Iduronate 2 sulfatase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Iduronate 2 sulfatase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Iduronate 2 sulfatase Antibody (CL488-66112) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 Iduronate 2 sulfatase antibody (CL488-66112, Clone: 1A3F9) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Iduronate 2 sulfatase Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-66112
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1A3F9
          Immunogen
          Iduronate 2 sulfatase fusion protein Ag19095
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MPPPRTGRG LLWLGLVLSS VCVALGSETQ ANSTTDALNV LLIIVDDLRP SLGCYGDKLV RSPNIDQLAS HSLLFQNAFA QQAVCAPSRV SFLTGRRPDT TRLYDFNSYW RVHAGNFSTI PQYFKENGYV TMSVGKVFHP GISSNHTDDS PYSWSFPPYH PSSEKYENTK TCRGPDGELH ANLLCPVDVL DVPEGTLPDK QSTEQAIQLL EKMKTSASPF FLAVGYHKPH IPFRYPKEFQ KLYPLENITL APDPEVPDGL PPVAYNPWMD IRQREDVQAL NISVPYGPIP VDFQEDQSST GFRLKTSSTR KYK
          Target Information
          Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.
          Bioinformatics
          Protein Aliases: Alpha-L-iduronate sulfate sulfatase; Iduronate 2-sulfatase; iduronate 2-sulfatase 14 kDa chain; iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; Idursulfase; unnamed protein product
          View more View less
          Gene Aliases: AW214631; ID2S; IDS; MPS2; SIDS
          View more View less
          UniProt ID: (Human) P22304, (Mouse) Q32KI7
          View more View less
          Entrez Gene ID: (Human) 3423, (Pig) 100512880, (Mouse) 15931, (Rat) 363513
          View more View less
          Function(s)
          iduronate-2-sulfatase activity calcium ion binding protein binding sulfuric ester hydrolase activity hydrolase activity metal ion binding catalytic activity molecular_function
          Process(es)
          metabolic process glycosaminoglycan catabolic process heparan sulfate proteoglycan catabolic process dermatan sulfate catabolic process carbohydrate derivative catabolic process biological_process
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline