    • Ciência Industrial e Aplicada
  • Novos produtos
  • Ofertas especiais
  • Fale Conosco
  • Pedido rápido
  • Documentos e certificados
Search button
          • Fale Conosco
          • Pedido rápido
          • Faça o login
            Faça o login
            Não tem uma conta? Criar Conta​
            • Conta
            • Status do pedido
            • Produtos Customizados & Projetos
          • Primary Antibodies ›
          • SMYD3 Antibodies
          Proteintech
          SMYD3 Monoclonal Antibody (1B5C10), CoraLite® Plus 488
          View all (16) SMYD3 antibodies
          Tech Support
          Cite SMYD3 Monoclonal Antibody (1B5C10), CoraLite® Plus 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SMYD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SMYD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SMYD3 Antibody (CL488-66330) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66330 (SMYD3 antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SMYD3 Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-66330
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1B5C10
          Immunogen
          SMYD3 fusion protein Ag2624
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MRCSQCRVA KYCSAKCQKK AWPDHKRECK CLKSCKPRYP PDSVRLLGRV VFKLMDGAPS ESEKLYSFYD LESNINKLTE DRKEGLRQLV MTFQHFMREE IQDASQLPPA FDLFEAFAKV ICNSFTICNA EMQEVGVGLY PSISLLNHSC DPNCSIVFNG PHLLLRAVRD IEVGEELTIC YLDMLMTSEE RRKQLRDQYC FECDCFRCQT QDKDADMLTG DEQVWKEVQE SLKKIEELKA HWKWEQVLAM CQAIISSNSE RLPDINIYQL KVLDCAMDAC INLGLLEEAL FYGTRTMEPY RIFFPGSHPV RGVQVMKVGK LQLHQGMFPQ AMKNLRLAFD IMRVTHGREH SLIEDLILLL EECDANIRAS
          Target Information
          SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
          Bioinformatics
          Protein Aliases: SET and MYND domain-containing protein 3; SET domain; unnamed protein product; zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1
          View more View less
          Gene Aliases: bA74P14.1; KMT3E; ZMYND1; ZNFN3A1
          View more View less
          UniProt ID: (Human) Q9H7B4
          View more View less
          Entrez Gene ID: (Human) 64754
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core binding RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding protein binding histone methyltransferase activity histone H4 methyltransferase activity histone H3K36 dimethyltransferase activity histone H3K4 trimethyltransferase activity
          Process(es)
          chromatin remodeling heart development negative regulation of transcription, DNA-templated
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline