    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SH3KBP1 Antibodies
          Proteintech
          CIN85 Monoclonal Antibody (3C12D2), CoraLite® Plus 488
          View all (17) SH3KBP1 antibodies
          Tech Support
          Cite CIN85 Monoclonal Antibody (3C12D2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CIN85 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CIN85 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CIN85 Antibody (CL488-66571) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66571 (CIN85 antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CIN85 Antibody in Western Blot (WB)
          Product Details
          CL488-66571
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C12D2
          Immunogen
          CIN85 fusion protein Ag2777
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MVEAIVEFD YQAQHDDELT ISVGEIITNI RKEDGGWWEG QINGRRGLFP DNFVREIKKE MKKDPLTNKA PEKPLHEVPS GNSLLSSETI LRTNKRGERR RRRCQVAFSY LPQNDDELEL KVGDIIEVVG EVEEGWWEGV LNGKTGMFPS NFIKELSGES DELGISQDEQ LSKSSLRETT GSESDGGDSS STKSEGANGT VATAAIQPKK VKGVGFGDIF KDKPIKLRPR SIEVENDFLP VEKTIGKKLP ATTATPDSSK TEMDSRTKSK DYCKVIFPYE AQNDDELTIK EGDIVTLINK DCIDVGWWEG ELNGRRGVFP DNFVKLLPP
          Target Information
          This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: c-Cbl-interacting protein; Cbl-interacting protein of 85 kDa; CD2-binding adaptor protein; CD2-binding protein 3; CD2BP3; cell growth-inhibiting gene 10 protein; HSB-1; Human Src family kinase-binding protein 1; MIG18; migration-inducing gene 18; Regulator of ubiquitous kinase; Ruk; SH3 domain-containing adapter protein; SH3 domain-containing kinase-binding protein 1; SH3-containing, expressed in tumorigenic astrocytes; SH3-domain kinase binding protein 1; Src family kinase-binding protein 1; src-related kinase binding protein-1; src-related kinase binding protein-1; HSB-1; unnamed protein product
          View more View less
          Gene Aliases: 1200007H22Rik; 1700125L08Rik; 5830464D22Rik; AGMX2; AI447724; CD2BP3; CIN85; GIG10; HSB-1; HSB1; IMD61; IN85; MIG18; Ruk; Seta; SH3KBP1
          View more View less
          UniProt ID: (Human) Q96B97, (Rat) Q925Q9, (Mouse) Q8R550
          View more View less
          Entrez Gene ID: (Human) 30011, (Rat) 84357, (Mouse) 58194
          View more View less
          Function(s)
          protein binding SH3 domain binding ubiquitin protein ligase binding
          Process(es)
          endocytosis apoptotic process cytoskeleton organization actin filament organization cell-cell signaling regulation of cell shape cell migration positive regulation of B cell activation regulation of apoptotic process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline