    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • CEP170B Antibodies
          Proteintech
          KIAA0284 Monoclonal Antibody (1D4B8), CoraLite® Plus 488
          View all (5) CEP170B antibodies
          Tech Support
          Cite KIAA0284 Monoclonal Antibody (1D4B8), CoraLite® Plus 488
          • Antibody Testing Data (1)
          KIAA0284 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          KIAA0284 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          KIAA0284 Antibody (CL488-66749) in Flow
          1x10^6 HeLa cells were intracellularly stained with 0.4 µg CoraLite® Plus 488 Anti-Human KIAA0284 (CL488-66749, Clone: 1D4B8) (red), or 0.4 µg Mouse IgG2b Isotype Control (CL488-66360-3, Clone: K11B8C4B5) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KIAA0284 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-66749
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D4B8
          Immunogen
          KIAA0284 fusion protein Ag26852
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LESPELSRA SSATFRPVIR GDRDESDDGG VAQRMALLQE FASRPLGAAP QAEHQGLPVP GSPGGQKWVS RWASLADSYS DPGLTEDGLG RRGGEPEGSL PVRMRRRLPQ LPSERADSPA GPESSRRSGP GPPELDSEQP SRLFGQEELD PDSLSDASGS DGGRGPEPGV EPQDSRRRSP QEGPTWSRGR RSPRAPGEPT PASFFIGDQN GDAVLSRKPL AAPGDGEGLG QTAQPSPPAR DGVYVSANGR MVIQLRPGRS PEPDGPAPAF LRQESFTKEP ASGPPAPGKP PHISSHPLLQ DLAATRAARM DFHSQDTHLI LKETETALAA LEARLLSNSV DAECEGGSTP RPPEDALS
          Target Information
          CEP170B plays a role in microtubule organization.
          Bioinformatics
          Protein Aliases: Centrosomal protein 170B; Centrosomal protein of 170 kDa protein B; Cep170-related; Cep170B
          View more View less
          Gene Aliases: CEP170B; CEP170R; FAM68C; KIAA0284
          View more View less
          UniProt ID: (Human) Q9Y4F5
          View more View less
          Entrez Gene ID: (Human) 283638
          View more View less
          Function(s)
          scaffold/adaptor protein
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0