    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • OGT Antibodies
          Proteintech
          OGT Monoclonal Antibody (2B2A6), CoraLite® Plus 488
          View all (20) OGT antibodies
          Tech Support
          Cite OGT Monoclonal Antibody (2B2A6), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          OGT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          OGT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          OGT Antibody (CL488-66823) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66938 (ZFP36 antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          OGT Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-66823
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:4,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2B2A6
          Immunogen
          OGT fusion protein Ag28402
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MASSVGNVAD STGLAELAHR EYQAGDFEAA ERHCMQLWRQ EPDNTGVLLL LSSIHFQCRR LDRSAHFSTL AIKQNPLLAE AYSNLGNVYK ERGQLQEAIE HYRHALRLKP DFIDGYINLA AALVAAGDME GAVQAYVSAL QYNPDLYCVR SDLGNLLKAL GRLEEAKACY LKAIETQPNF AVAWSNLGCV FNAQGEIWLA IHHFEKAVTL DPNFLDAYIN LGNVLKEARI FDRAVAAYLR ALSLSPNHAV VHGNLACVYY EQGLIDLAID TYRRAIELQP HFPDAYCNLA NALKEKGSVA EAEDCYNTAL RLCPTHADSL NNLANIKREQ GNIEEAVRLY RKALEVFPEF AAAHSNLASV LQQQGKLQEA LMHYKEAIRI SPTFADAYSN MGNTLKEMQD
          Target Information
          This gene encodes a glycosyltransferase that catalyzes the addition of a single N-acetylglucosamine in O-glycosidic linkage to serine or threonine residues. Since both phosphorylation and glycosylation compete for similar serine or threonine residues, the two processes may compete for sites, or they may alter the substrate specificity of nearby sites by steric or electrostatic effects. The protein contains multiple tetratricopeptide repeats that are required for optimal recognition of substrates. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: FLJ23071; MGC22921; O linked N-acetylglucosamine transferase; o-glcnac transferase; O-GlcNAc transferase subunit p110; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; OGT; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N- acetylglucosaminyl transferase; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase
          View more View less
          Gene Aliases: 1110038P24Rik; 4831420N21Rik; AI115525; OGT; Ogtl
          View more View less
          UniProt ID: (Pig) Q27HV0, (Mouse) Q8CGY8, (Rat) P56558
          View more View less
          Entrez Gene ID: (Pig) 664652, (Mouse) 108155, (Rat) 26295
          View more View less
          Function(s)
          phosphatidylinositol-3,4,5-trisphosphate binding protein N-acetylglucosaminyltransferase activity histone acetyltransferase activity (H4-K5 specific) histone acetyltransferase activity (H4-K8 specific) histone acetyltransferase activity (H4-K16 specific) protein O-GlcNAc transferase activity catalytic activity protein binding enzyme activator activity N-acetyltransferase activity transcription factor binding lipid binding transferase activity transferase activity, transferring glycosyl groups protein domain specific binding peptide binding monosaccharide binding protein modifying enzyme
          Process(es)
          regulation of glycolytic process regulation of gluconeogenesis protein O-linked glycosylation apoptotic process protein processing negative regulation of protein ubiquitination circadian regulation of gene expression histone H4-K5 acetylation histone H4-K8 acetylation histone H4-K16 acetylation positive regulation of proteolysis positive regulation of transcription from RNA polymerase II promoter phosphatidylinositol-mediated signaling positive regulation of histone H3-K27 methylation histone H3-K4 trimethylation negative regulation of protein phosphorylation positive regulation of protein phosphorylation glucosamine metabolic process positive regulation of gene expression negative regulation of peptidyl-threonine phosphorylation chromatin modification positive regulation of granulocyte differentiation response to insulin negative regulation of peptidyl-serine phosphorylation regulation of Rac protein signal transduction positive regulation of catalytic activity positive regulation of cell size regulation of insulin receptor signaling pathway intracellular distribution of mitochondria rhythmic process positive regulation of histone H3-K4 methylation negative regulation of cell death protein homotrimerization protein heterotrimerization cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to glucose stimulus negative regulation of protein targeting to membrane regulation of gluconeogenesis involved in cellular glucose homeostasis negative regulation of cellular response to hypoxia positive regulation of protein localization to nucleus positive regulation of reactive oxygen species biosynthetic process forebrain development cellular response to insulin stimulus cellular response to toxic substance
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline