    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • AIMP2 Antibodies
          Proteintech
          JTV1 Monoclonal Antibody (1D6B5), CoraLite® Plus 488
          View all (11) AIMP2 antibodies
          Tech Support
          Cite JTV1 Monoclonal Antibody (1D6B5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          JTV1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          JTV1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          JTV1 Antibody (CL488-66848) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-66848 (JTV1 antibody) at dilution of 1:800 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          JTV1 Antibody in Western Blot (WB)
          JTV1 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-66848
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D6B5
          Immunogen
          JTV1 fusion protein Ag0675
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MPMYQVKPY HGGGAPLRVE LPTCMYRLPN VHGRSYGPAP GAGHVQEESN LSLQALESRQ DDILKRLYEL KAAVDGLSKM IQTPDADLDV TNIIQADEPT TLTTNALDLN SVLGKDYGAL KDIVINANPA SPPLSLLVLH RLLCEHFRVL STVHTHSSVK SVPENLLKCF GEQNKKQPRQ DYQLGFTLIW KNVPKTQMKF S
          Target Information
          JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.
          Bioinformatics
          Protein Aliases: Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV1; Multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; Protein JTV-1; unnamed protein product
          View more View less
          Gene Aliases: AA407136; AA407225; AIMP2; Aimp2(p38); HLD17; JTV-1; JTV1; P38; PRO0992
          View more View less
          UniProt ID: (Human) Q13155, (Rat) Q32PX2, (Mouse) Q8R010
          View more View less
          Entrez Gene ID: (Human) 7965, (Rat) 288480, (Mouse) 231872
          View more View less
          Function(s)
          protein binding protein binding, bridging binding, bridging scaffold/adaptor protein
          Process(es)
          translation apoptotic process negative regulation of cell proliferation protein ubiquitination cell differentiation positive regulation of protein ubiquitination positive regulation of neuron apoptotic process Type II pneumocyte differentiation protein-containing complex assembly positive regulation of neuron death multicellular organism development
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline