    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • DRG2 Antibodies
          Proteintech
          DRG2 Monoclonal Antibody (3A3E1), CoraLite® Plus 488
          View all (3) DRG2 antibodies
          Tech Support
          Cite DRG2 Monoclonal Antibody (3A3E1), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DRG2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DRG2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DRG2 Antibody (CL488-67164) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed U2OS cells using CoraLite® Plus 488 DRG2 antibody (CL488-67164, Clone: 3A3E1) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DRG2 Antibody in Immunocytochemistry (ICC/IF)
          DRG2 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-67164
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          1x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3A3E1
          Immunogen
          DRG2 fusion protein Ag6878
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MGILEKISE IEKEIARTQK NKATEYHLGL LKAKLAKYRA QLLEPSKSAS SKGEGFDVMK SGDARVALIG FPSVGKSTFL SLMTSTASEA ASYEFTTLTC IPGVIEYKGA NIQLLDLPGI IEGAAQGKGR GRQVIAVART ADVIIMMLDA TKGEVQRSLL EKELESVGIR LNKHKPNIYF KPKKGGGISF NSTVMLTQCS EKLVQLILHE YKIFNAEVLF REDCSPDEFI DVIVGNRVYM PCLYVYNKID QISMEEVDRL ARKPNSVVIS CGMKLNLDYL LEMLWEYLAL TCIYTKKRGQ RPDFTDAIIL RKGASVEHVC HRIHRSLASQ FKYALVWGTS TKYSPQRVGL THTMEHEDVI QIVKK
          Target Information
          3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
          Bioinformatics
          Protein Aliases: developmentally regulated GTP binding protein 2; developmentally regulated GTP-binding protein 2; Developmentally-regulated GTP-binding protein 2; DRG-2; TRAFAC GTPase DRG2; Translation factor GTPase DRG2; unnamed protein product
          View more View less
          Gene Aliases: AI255295; AI461605; DRG2; RGD1562380
          View more View less
          UniProt ID: (Human) P55039, (Mouse) Q9QXB9
          View more View less
          Entrez Gene ID: (Pig) 110256132, (Human) 1819, (Rat) 497915, (Mouse) 13495
          View more View less
          Function(s)
          nucleotide binding RNA binding GTPase activity protein binding GTP binding hydrolase activity metal ion binding
          Process(es)
          biological_process cytoplasmic translation signal transduction
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline