    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Munc13-4 Antibodies
          Proteintech
          UNC13D Monoclonal Antibody (1C5C1), CoraLite® Plus 488
          View all (13) Munc13-4 antibodies
          Tech Support
          Cite UNC13D Monoclonal Antibody (1C5C1), CoraLite® Plus 488
          • Antibody Testing Data (1)
          UNC13D Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          UNC13D Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          UNC13D Antibody (CL488-67193) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL488-67193 (UNC13D antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UNC13D Antibody in Western Blot (WB)
          Product Details
          CL488-67193
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1C5C1
          Immunogen
          UNC13D fusion protein Ag10497
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MATLLSHPQ QRPPFLRQAI KIRRRRVRDL QDPPPQMAPE IQPPSHHFSP EQRALLYEDA LYTVLHRLGH PEPNHVTEAS ELLRYLQEAF HVEPEEHQQT LQRVRELEKP IFCLKATVKQ AKGILGKDVS GFSDPYCLLG IEQGVGVPGG SPGSRHRQKA VVRHTIPEEE THRTQVITQT LNPVWDETFI LEFEDITNAS FHLDMWDLDT VESVRQKLGE LTDLHGLRRI FKEARKDKGQ DDFLGNVVLR LQDLRCREDQ WYPLEPRTET YPDRGQCHLQ FQLIHKRRAT SASRSQPSYT VHLHLLQQLV SHEVTQHEAG STSWDGSL
          Target Information
          This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
          Bioinformatics
          Protein Aliases: munc 13-4; Munc13-4; Protein unc-13 homolog D
          View more View less
          Gene Aliases: FHL3; HLH3; HPLH3; Munc13-4; UNC13D
          View more View less
          UniProt ID: (Human) Q70J99
          View more View less
          Entrez Gene ID: (Human) 201294
          View more View less
          Function(s)
          protein binding Rab GTPase binding
          Process(es)
          granuloma formation germinal center formation phagocytosis regulation of mast cell degranulation natural killer cell degranulation positive regulation of exocytosis defense response to virus positive regulation of substrate adhesion-dependent cell spreading positive regulation of regulated secretory pathway
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0