Immunogen sequence: MTECFLPPT SSPSEHRRVE HGSGLTRTPS SEEISPTKFP GLYRTGEPSP PHDILHEPPD VVSDDEKDHG KKKGKFKKKE KRTEGYAAFQ EDSSGDEAES PSKMKRSKGI HVFKKPSFSK KKEKDFKIKE KPKEEKHKEE KHKEEKHKEK KSKDLTAADV VKQWKEKKKK KKPIQEPEVP QIDVPNLKPI FGIPLADAVE RTMMYDGIRL PAVFRECIDY VEKYGMKCEG IYRVSGIKSK VDELKAAYDR EESTNLEDYE PNTVASLLKQ YLRDLPENLL TKELMPRFEE ACGRTTETEK VQEFQRLLKE LPECNYLLIS WLIVHMDHVI AKELETKMNI QNISIVLSPT VQ
Can specifically activate hydrolysis of GTP bound to RAC1 and CDC42, but not RALA. Mediates ATP-dependent transport of S-(2,4-dinitrophenyl)-glutathione (DNP-SG) and doxorubicin (DOX) and is the major ATP-dependent transporter of glutathione conjugates of electrophiles (GS-E) and DOX in erythrocytes. Can catalyze transport of glutathione conjugates and xenobiotics, and may contribute to the multidrug resistance phenomenon. Serves as a scaffold protein that brings together proteins forming an endocytotic complex during interphase and also with CDC2 to switch off endocytosis, One of its substrates would be EPN1/Epsin.
Protein Aliases: 76 kDa Ral-interacting protein; Dinitrophenyl S-glutathione ATPase; DNP-SG ATPase; Ral-interacting protein 1; RalA-binding protein 1; RalBP1; RIP; unnamed protein product
Gene Aliases: RALBP1; RIP1; RLIP; RLIP1; RLIP76
UniProt ID: (Human) Q15311, (Mouse) Q62172
Entrez Gene ID: (Human) 10928, (Mouse) 19765