    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • UGDH Antibodies
          Proteintech
          UGDH Monoclonal Antibody (1E2E1), CoraLite® Plus 488
          View all (11) UGDH antibodies
          Tech Support
          Cite UGDH Monoclonal Antibody (1E2E1), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          UGDH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UGDH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          UGDH Antibody (CL488-67360) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite® Plus 488 UGDH antibody (CL488-67360, Clone: 1E2E1) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UGDH Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-67360
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1E2E1
          Immunogen
          UGDH fusion protein Ag29747
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MFEIKKICC IGAGYVGGPT CSVIAHMCPE IRVTVVDVNE SRINAWNSPT LPIYEPGLKE VVESCRGKNL FFSTNIDDAI KEADLVFISV NTPTKTYGMG KGRAADLKYI EACARRIVQN SNGYKIVTEK STVPVRAAES IRRIFDANTK PNLNLQVLSN PEFLAEGTAI KDLKNPDRVL IGGDETPEGQ RAVQALCAVY EHWVPREKIL TTNTWSSELS KLAANAFLAQ RISSINSISA LCEATGADVE EVATAIGMDQ RIGNKFLKAS VGFGGSCFQK DVLNLVYLCE ALNLPEVARY WQ
          Target Information
          The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia.
          Bioinformatics
          Protein Aliases: UDP UGD; UDP-Glc dehydrogenase; UDP-GlcDH; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogeanse; UDP-glucose dehydrogenase; UDPGDH; UDPGDH; NAD+-linked oxidoreductase; unnamed protein product; uridine diphospho-glucose dehydrogenase
          View more View less
          Gene Aliases: DEE84; EIEE84; GDH; UDP-GlcDH; UDPGDH; UGD; UGDH
          View more View less
          UniProt ID: (Human) O60701, (Rat) O70199, (Mouse) O70475
          View more View less
          Entrez Gene ID: (Pig) 100627727, (Human) 7358, (Rat) 83472, (Mouse) 22235
          View more View less
          Function(s)
          catalytic activity UDP-glucose 6-dehydrogenase activity oxidoreductase activity oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor identical protein binding NAD binding dehydrogenase
          Process(es)
          gastrulation with mouth forming second glycosaminoglycan biosynthetic process UDP-glucuronate biosynthetic process heparan sulfate proteoglycan biosynthetic process protein hexamerization neuron development chondroitin sulfate proteoglycan biosynthetic process oxidation-reduction process metabolic process
          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          Korea flag icon
          Korea
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)
          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)
          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline