    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PSMG3 Antibodies
          Proteintech
          PSMG3 Monoclonal Antibody (2H9C3), CoraLite® Plus 488
          View all (2) PSMG3 antibodies
          Tech Support
          Cite PSMG3 Monoclonal Antibody (2H9C3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          PSMG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PSMG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PSMG3 Antibody (CL488-67466) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using CoraLite® Plus 488 PSMG3 antibody (CL488-67466, Clone: 2H9C3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PSMG3 Antibody in Immunocytochemistry (ICC/IF)
          PSMG3 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-67466
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2H9C3
          Immunogen
          PSMG3 fusion protein Ag8384
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEDTPLVIS KQKTEVVCGV PTQVVCTAFS SHILVVVTQF GKMGTLVSLE PSSVASDVSK PVLTTKVLLG QDEPLIHVFA KNLVAFVSQE AGNRAVLLAV AVKDKSMEGL KALREVIRVC QVW
          Target Information
          PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.
          Bioinformatics
          Protein Aliases: hPAC3; PAC-3; Pba3; proteasome (prosome, macropain) assembly chaperone 3; proteasome assembling chaperone 3; Proteasome assembly chaperone 3; Proteasome chaperone homolog 3
          View more View less
          Gene Aliases: C7orf48; PAC3; Pba3; PSMG3
          View more View less
          UniProt ID: (Human) Q9BT73
          View more View less
          Entrez Gene ID: (Human) 84262
          View more View less
          Function(s)
          protein binding macromolecular complex binding binding, bridging
          Process(es)
          proteasome assembly chaperone-mediated protein complex assembly
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline