Immunogen sequence: MIKLFSLKQQ KKEEESAGGT KGSSKKASAA QLRIQKDINE LNLPKTCDIS FSDPDDLLNF KLVICPDEGF YKSGKFVFSF KVGQGYPHDP PKVKCETMVY HPNIDLEGNV CLNILREDWK PVLTINSIIY GLQYLFLEPN PEDPLNKEAA EVLQNNRRLF EQNVQRSMRG GYIGSTYFER CLK
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.
Protein Aliases: epididymis secretory sperm binding protein; NEDD8 carrier protein; NEDD8 protein ligase; NEDD8-conjugating enzyme Ubc12; UBC12 homolog, yeast; ubiquitin C, related sequence 2; ubiquitin carrier protein M; ubiquitin conjugating enzyme E2M; Ubiquitin-conjugating enzyme E2 M; ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast); ubiquitin-protein ligase M; yeast UBC12 homolog
Gene Aliases: 2510040H03Rik; hUbc12; UBC-RS2; UBC12; UBE2M
UniProt ID: (Human) P61081, (Mouse) P61082
Entrez Gene ID: (Human) 9040, (Mouse) 22192, (Rat) 361509