    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PIK3R2 Antibodies
          Proteintech
          PI3 Kinase p85 Beta Monoclonal Antibody (2B3B6), CoraLite® Plus 488
          View all (23) PIK3R2 antibodies
          Tech Support
          Cite PI3 Kinase p85 Beta Monoclonal Antibody (2B3B6), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          PI3 Kinase p85 Beta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PI3 Kinase p85 Beta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PI3 Kinase p85 Beta Antibody (CL488-67644) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite® Plus 488 PI3 Kinase p85 Beta antibody (CL488-67644, Clone: 2B3B6) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PI3 Kinase p85 Beta Antibody in Immunocytochemistry (ICC/IF)
          PI3 Kinase p85 Beta Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-67644
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2B3B6
          Immunogen
          PI3 Kinase p85 Beta fusion protein Ag14369
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: YRHESLAQY NAKLDTRLLY PVSKYQQDQI VKEDSVEAVG AQLKVYHQQY QDKSREYDQL YEEYTRTSQE LQMKRTAIEA FNETIKIFEE QGQTQEKCSK EYLERFRREG NEKEMQRILL NSERLKSRIA EIHESRTKLE QQLRAQASDN REIDKRMNSL KPDLMQLRKI RDQYLVWLTQ KGARQKKINE WLGIKNETED QYALMEDEDD LPHHEERTWY VGKINRTQAE EMLSGKRDGT FLIRESSQRG CYACSVVVDG DTKHCVIYRT ATGFGFAEPY NLYGSLKELV LHYQHASLVQ HNDALTVTLA HPVRAPGPGP PPAAR
          Target Information
          Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.
          Bioinformatics
          Protein Aliases: Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; Phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 2 (beta); PI3-kinase p85 subunit beta; PI3-kinase regulatory subunit beta; PI3-kinase subunit p85-beta; PI3K; PI3K regulatory subunit beta; ptdIns-3-kinase p85-beta; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit p85-beta
          View more View less
          Gene Aliases: MPPH; MPPH1; p85; p85-BETA; P85B; p85beta; PIK3R2
          View more View less
          UniProt ID: (Human) O00459, (Mouse) O08908
          View more View less
          Entrez Gene ID: (Human) 5296, (Mouse) 18709, (Rat) 29741
          View more View less
          Function(s)
          phosphotyrosine binding GTPase activator activity protein binding protein phosphatase binding receptor tyrosine kinase binding phosphatidylinositol 3-kinase regulatory subunit binding 1-phosphatidylinositol-3-kinase regulator activity protein heterodimerization activity transcription factor binding 1-phosphatidylinositol-3-kinase activity phosphatidylinositol 3-kinase regulator activity kinase modulator
          Process(es)
          cellular glucose homeostasis immune response signal transduction insulin receptor signaling pathway regulation of autophagy protein transport B cell differentiation T cell differentiation regulation of actin filament polymerization cellular response to insulin stimulus response to endoplasmic reticulum stress positive regulation of protein import into nucleus negative regulation of MAPK cascade protein kinase B signaling positive regulation of cell adhesion positive regulation of transcription from RNA polymerase II promoter regulation of stress fiber assembly regulation of protein localization to plasma membrane transport phosphatidylinositol 3-kinase signaling regulation of phosphorylation positive regulation of transcription factor import into nucleus regulation of phosphatidylinositol 3-kinase activity phosphatidylinositol phosphorylation regulation of establishment of protein localization to plasma membrane phosphorylation phosphatidylinositol-3-phosphate biosynthetic process
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0