FIGURE: 1 / 1
Immunogen sequence: MELHYLAKKS NQADLCDARD WSSRGLPGDQ ADTAATRAAL CCQKQCASTP RATEMEGSKL SSSPASPSSS LQNSTLQPDA FPPGLLHSGN NQITAERKVC NCCSQELETS FTYVDKNINL EQRNRSSPSA KGHNHPGELG WENPNEWSQE AAISLISEEE DDTSSEATSS GKSIDYGFIS AILFLVT
Protein Aliases: Transmembrane protein 74; unnamed protein product
Gene Aliases: NET36; TMEM74
UniProt ID: (Human) Q96NL1
Entrez Gene ID: (Human) 157753